Skip to Content
Merck
All Photos(1)

Key Documents

MAB1892

Sigma-Aldrich

Anti-Fibronectin Antibody, gelatin binding domain, clone IST-10

clone IST-10, Chemicon®, from mouse

Synonym(s):

Anti-CIG, Anti-ED-B, Anti-FINC, Anti-FN, Anti-FNZ, Anti-GFND, Anti-GFND2, Anti-LETS, Anti-MSF, Anti-SMDCF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IST-10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)
mouse ... Fn1(14268)

Specificity

Recognizes an epitope contained within the gelatin binding domain of human fibronectin.

Immunogen

Epitope: gelatin binding domain

Application

Anti-Fibronectin Antibody, gelatin binding domain, clone IST-10 is an antibody against Fibronectin for use in WB, IH.
Immunohistochemistry

Immunoblotting: non-reducing conditions

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Purified immunoglobulin in 0.02M phosphate buffer, 0.25M sodium chloride, 0.1% sodium azide, pH 7.6

Storage and Stability

Maintain between 2 and 8°C .

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jane Sottile et al.
American journal of physiology. Cell physiology, 293(6), C1934-C1946 (2007-10-12)
Communication between cells and the extracellular matrix (ECM) is critical for regulation of cell growth, survival, migration, and differentiation. Remodeling of the ECM can occur under normal physiological conditions, as a result of tissue injury, and in certain pathological conditions.
Definition of the native and denatured type II collagen binding site for fibronectin using a recombinant collagen system.
An, B; Abbonante, V; Yigit, S; Balduini, A; Kaplan, DL; Brodsky, B
The Journal of Biological Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service