Skip to Content
Merck
All Photos(1)

Key Documents

IPFL10100

Millipore

Immobilon® -FL PVDF Membrane

10 sheets, 10 cm x 10 cm, 0.45 µm pore size, transfer membrane with low background fluorescence

Synonym(s):

Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22
material:
PVDF membrane
plain filter
white filter
detection method:
chemiluminescent
colorimetric
fluorometric
pore size:
0.45 μm pore size
compatibility:
for use with Amido black
for use with CPTS
for use with Coomassie brilliant blue
for use with Ponceau-S red

Product Name

Immobilon®-FL PVDF Membrane, 10 sheets, 10 cm x 10 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane with low background fluorescence for Western blotting. Compatible with visible and infrared fluorescent probes.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

10 cm × 10 cm

pore size

0.45 μm pore size

capacity

155 μg/cm2 adsorption capacity (insulin)
205 μg/cm2 adsorption capacity (BSA)
300 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Coomassie brilliant blue
for use with Ponceau-S red

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

General description

The Immobilon®-FL transfer membrane is hydrophobic polyvinylidene fluoride (PVDF) microporous membrane used for the transfer of proteins from a variety of gel matrices. This transfer membrane offers a uniformly controlled pore structure with a high binding capacity for biomolecules. The membrane displays very low autofluorescence across a wide range of excitation/emission wavelengths. The Immobilon®-FL membrane has a nominal pore size of 0.45 μm and offers optimal blotting for proteins with molecular weights greater than 20,000 Daltons.

Application

Immobilon®-FL membrane has been used in western blotting. It is also suitable for use in standard chemiluminescent or chromogenic detection.

Features and Benefits

Immobilon®-FL Membrane:
  • The first transfer membrane designed for fluorescence applications Extremely low background improves the sensitivity of all fluorescence detection protocols
  • Compatible with all commonly used fluorescent probes at all excitation and emission wavelengths
  • compatible with standard blocking agents and buffers
  • Ideal for multiplexing and chemifluorescence detections

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yerim Lee et al.
eLife, 8 (2019-11-02)
Membrane nanodomains have been implicated in Ras signaling, but what these domains are and how they interact with Ras remain obscure. Here, using single particle tracking with photoactivated localization microscopy (spt-PALM) and detailed trajectory analysis, we show that distinct membrane
Jessica E Chukwu et al.
Scientific reports, 8(1), 6209-6209 (2018-04-20)
Tau antibodies have shown therapeutic potential for Alzheimer's disease and several are in clinical trials. As a microtubule-associated protein, tau relies on dynamic phosphorylation for its normal functions. In tauopathies, it becomes hyperphosphorylated and aggregates into toxic assemblies, which collectively
Maria Consiglia Trotta et al.
PloS one, 14(1), e0211005-e0211005 (2019-01-19)
The role of the hydroxycarboxylic acid receptor 2 (HCA2) in the retinal damage induced by diabetes has never been explored. In this context, the present study highlights an upregulation of retinal HCA2 receptors in diabetic C57BL6J mice. Moreover, we illustrate
Shane M Bemiller et al.
Journal of neuroinflammation, 15(1), 278-278 (2018-09-27)
Fractalkine (CX3CL1) and its receptor (CX3CR1) play an important role in regulating microglial function. We have previously shown that Cx3cr1 deficiency exacerbated tau pathology and led to cognitive impairment. However, it is still unclear if the chemokine domain of the
Rosa Maisto et al.
Frontiers in pharmacology, 11, 235-235 (2020-03-27)
Stimulation of retinal photoreceptors with elevated glucose concentration (30 mM) for 96 h, served as diabetic retinopathy in vitro model to study Resolvin D1 (50 nM) effects on neovascularization. VEGF and anti-angiogenic miR-20a-3p, miR-20a-5p, miR-106a-5p, and miR-20b expression was assessed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service