Skip to Content
Merck
All Photos(1)

Documents

IPFL00010

Millipore

Immobilon® -FL PVDF Membrane

1 roll, 27 cm x 3.75 m, 0.45 µm pore size, transfer membrane with low background fluorescence

Synonym(s):

Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

product name

Immobilon®-FL PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane with low background fluorescence for Western blotting. Compatible with visible and infrared fluorescent probes.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

27 cm × 3.75 m

pore size

0.45 μm pore size

capacity

155 μg/cm2 adsorption capacity (insulin)
205 μg/cm2 adsorption capacity (BSA)
300 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with All fluorescence dyes
for use with Amido black
for use with CPTS
for use with Coomassie brilliant blue
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

General description

The Immobilon®-FL transfer membrane is a polyvinylidene fluoride (PVDF) microporous membrane with a nominal pore size of 0.45 μm. It is designed for binding proteins transferred from a variety of gel matrices. The Immobilon®-FL transfer membrane is hydrophobic and offers a uniformly controlled pore structure with a high binding capacity for biomolecules. The membrane offers optimal blotting for proteins with molecular weights greater than 20,000. The Immobilon®-FL membrane displays very low autofluorescence across a wide range of excitation/emission wavelengths. This property makes it ideal for blotting applications involving fluorescence-based immunodetection. Immobilon®-FL membrane can also be used for standard chemiluminescent or chromogenic detection.

Application

Immobilon-FL PVDF Membrane has been used in western blotting.

Features and Benefits

Immobilon-FL Membrane:
  • The first transfer membrane optimized for fluorescence applications; Extremely low background enhances the sensitivity of all fluorescence detection protocols
  • Compatible with all commonly used fluorescent probes at a wide range of excitation/emission wavelengths
  • Best for multiplexing and chemifluorescence detections
  • compatible with standard blocking agents and buffers

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangchuan Wang et al.
Biological research, 52(1), 60-60 (2019-12-19)
Recent studies have confirmed that RASAL1 has an antitumor effect in many cancers, but its functional role and the molecular mechanism underlying in colon cancer has not been investigated. We collected human colon cancer tissues and adjacent non-tumor tissues, human
S A Misek et al.
Oncogene, 39(7), 1466-1483 (2019-10-30)
Over half of cutaneous melanoma tumors have BRAFV600E/K mutations. Acquired resistance to BRAF inhibitors (BRAFi) remains a major hurdle in attaining durable therapeutic responses. In this study we demonstrate that ~50-60% of melanoma cell lines with vemurafenib resistance acquired in
Daniel Del Valle Morales et al.
Nucleic acids research, 48(7), 3806-3815 (2020-01-31)
Cap homeostasis is the cyclical process of decapping and recapping that maintains the translation and stability of a subset of the transcriptome. Previous work showed levels of some recapping targets decline following transient expression of an inactive form of RNMT
David J Ferland et al.
PloS one, 15(2), e0229251-e0229251 (2020-02-25)
Since chemerin's identification as an adipokine, it has been associated with a number of human diseases including diabetes and obesity. However, the basic scientific foundation for these clinical determinations is still lacking. Fibroblastic mouse 3T3 cells are unable to develop
Mark Jackman et al.
The Journal of cell biology, 219(6) (2020-04-03)
How the cell rapidly and completely reorganizes its architecture when it divides is a problem that has fascinated researchers for almost 150 yr. We now know that the core regulatory machinery is highly conserved in eukaryotes, but how these multiple

Protocols

This page shows and discusses three protocols for stripping and reprobing a western blot membrane.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service