Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
sample type cell culture supernatant
sample type serum
Product Name
Human IL-8 Conferma® ELISA, quantification of Human IL-8 in serum, plasma, or culture media samples
Quality Segment
species reactivity
human
parameter
50 μL sample volume (4hr assay)
technique(s)
ELISA: suitable
input
sample type plasma (K2 EDTA)
sample type cell culture supernatant
sample type serum
UniProt accession no.
application(s)
research use
assay range
accuracy: 77-102 (plasma)%
linearity: 105 (plasma)%, inter-assay cv: <4.7%
intra-assay cv: <4.0%
standard curve range: 0.82-200 pg/mL
accuracy: 70-105 (serum)%
linearity: 94-106 (serum)%
detection method
colorimetric (450nm/590nm)
storage temp.
2-8°C
Gene Information
human ... CXCL8(3576)
General description
IL-8(6-77) has a 5-10-fold higher activity on neutrophil activation, IL-8(5-77) has increased activity on neutrophil activation and IL-8(7-77) has a higher affinity to receptors CXCR1 and CXCR2 as compared to IL-8(1-77), respectively.
This is a 96 well format sandwich ELISA which uses our “Conferma” process to ensure a high degree of lot to lot consistency. The process involves the physiochemical characterization of the ELISAs critical reagents lot to lot, with finished kits being evaluated using a sample library to bridge between lots.
Application
Analytes available:
- IL-8
Research Category:
- Cytokines
- Inflammation & Immunology
- Immune Response
- Metabolic Disease
Analysis Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information human ... CXCL8(3576) | Gene Information human ... IL6(3569) | Gene Information - | Gene Information human ... IL1B(3553) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| input sample type plasma (K2 EDTA) | input sample type serum | input sample type plasma (K2 EDTA) | input sample type urine |
| assay range accuracy: 77-102 (plasma)% | assay range - | assay range accuracy: 101-107% | assay range - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level - |
| detection method colorimetric (450nm/590nm) | detection method colorimetric (450nm/590nm) | detection method colorimetric (450nm/590nm) | detection method colorimetric |
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



