Skip to Content
Merck
All Photos(1)

Key Documents

EZHADP

Millipore

Human Adiponectin ELISA Kit

measures Adiponectin in 20 μL serum, plasma or cell culture supernatent

Synonym(s):

Adiponectin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

product name

Human Adiponectin ELISA, This Human Adiponectin ELISA is used to measure & quantify Adiponectin levels in Metabolism & Endocrine research.

Quality Level

species reactivity

human

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr assay)

assay range

accuracy: 92-102%
intra-assay cv: 1.0-7.4
linearity: 81-100%
sensitivity: 1.5 ng/mL
standard curve range: 1.5-100 ng/mL
(adiponection)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma (K2 EDTA)

NCBI accession no.

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... ADIPOQ(9370)

General description

Adiponectin is identified as a 30 kDa adipokine that is secreted from the adipose tissue into circulating blood and influences systemic metabolism.

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders
Room Temperature, 3 hour assay using 20 µL sample size (serum/plasma: 1:500 dilution)
Used to detect/quantify: Adiponectin

Linkage

Replaces: CYT350

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ravi Retnakaran et al.
Journal of the American Heart Association, 8(21), e014231-e014231 (2019-10-28)
Background The 1-hour glucose challenge test (GCT) is routinely performed in pregnancy to screen for gestational diabetes mellitus. Remarkably, it has recently emerged that the GCT can also predict a woman's future risk of cardiovascular disease, although the mechanistic basis
Effect of antipsychotics on peptides involved in energy balance in drug-naive psychotic patients after 1 year of treatment.
Rocio Perez-Iglesias,Jose Luis Vazquez-Barquero,Jose Antonio Amado,Ana Berja et al.
Journal of Clinical Psychopharmacology null
Effects of acute hyperinsulinaemia on total and high-molecular-weight adiponectin concentration in metabolically healthy but obese postmenopausal women: a Montreal-Ottawa New Emerging Team (MONET) study.
Elisha B, Karelis AD, Imbeault P, Rabasa-Lhoret R
Diabetes and Metabolism null
Adiponectin changes in HCV-Genotype 4: relation to liver histology and response to treatment.
M Derbala, N Rizk, S Al-Kaabi, A Amer, F Shebl, A Al Marri, I Aigha, D Alyaesi, H Mohamed et al.
Journal of Viral Hepatitis null
Adiponectin levels and its association with hyperglycaemia in adult Filipino participants in the 2003--04 National Nutrition and Health Survey.
Paz-Pacheco E, Lim-Abrahan MA, Sy RA, Jasul GV, Sison CM, Laurel AF
Diabetes & Vascular Disease Research : Official Journal of the International Society of Diabetes and Vascular Disease null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service