Skip to Content
Merck

EZFREAD01DK

EZ-Fluo Reader

Contains EZ-Fluo reader, power cord for Denmark

Synonym(s):

Detection instrument, EZ-Fluo Reader kit (DK), For rapid detection of microbial contamination in filterable samples, suitable for bioburden testing

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.79
UNSPSC Code:
41116129
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

100 V / 240 V, 50 - 60 Hz

Quality Segment

manufacturer/tradename

EZ-Fluo

parameter

4.4 kg at 15 - 40 °C

H × W × L

12.5 cm × 14.2 cm × 24.9 cm

application(s)

environmental
food and beverages
pharmaceutical

compatibility

EZ-Fluo Rapid Microbial Detection System

General description

The EZ-Fluo reader is part of the EZ-Fluo Rapid detection system. It is a fluorescent-based technology designed for fast and quantitative detection of microorganisms over a broad range of filterable samples. Fluorescent micro-colonies can be counted directly through the viewing window of the reader. The reader is supplied with the power cord according to Denmark region.

Application

EZ-Fluo reader is a Rapid & non-destructive system for microbial detection in filterable samples

Legal Information

EZ-FLUO is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under EZ-Fluo Reader




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library