Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EZ-Fluo
™
Reader
Products
Product Category
instruments (11)
Brand
Millipore (11)
Agency
Length (cm)
Length (in)
Application
Housing Material
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
EZFREAD01EU
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for European countries
Expand
View Pricing
EZFREAD01UK
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for United Kingdom
Expand
View Pricing
EZFREAD01BR
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for Brazil
Expand
View Pricing
EZFREAD01IN
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for India
Expand
View Pricing
EZFREAD01AS
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for Australia, New Zealand, and Argentina
Expand
View Pricing
EZFREAD01IT
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for Italy
Expand
View Pricing
EZFREAD01SZ
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for Switzerland
Expand
View Pricing
EZFREAD01CN
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for China
Expand
View Pricing
EZFREAD01DK
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for Denmark
Expand
View Pricing
EZFREAD01JP
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for Japan
Expand
View Pricing
EZFREAD01MX
EZ-Fluo
™
Reader,
Contains EZ-Fluo
™
reader, power cord for Mexico
Expand
View Pricing