DAWP29325
MF-Millipore Membrane
pore size 0.65 μm, filter diam. 293 mm, hydrophilic
Synonym(s):
MF-Millipore Membrane, mixed cellulose esters, Hydrophilic, 0.65 μm, 293 mm, white, plain
About This Item
material
mixed cellulose esters (MCE)
plain filter
white filter
Quality Level
feature
hydrophilic
manufacturer/tradename
MF-Millipore™
parameter
140 mL/min-cm2 water flow rate
75 °C max. temp.
9 L/min-cm2 air flow rate
filter diam.
293 mm
thickness
180 μm
gravimetric extractables
3%
refractive index
n/D 1.51
matrix
MF-Millipore™
pore size
0.65 μm pore size
81 % porosity
bubble point
≥1.17 bar, air with water at 23 °C
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
- Organism Retention: Microorganism
- Mode of Action: Filtration (size exclusion)
- Application: BioProcessing
- Intended Use: Reduction or removal of microorganism/bioburden
- Instructions for Use: Please refer first page of Visual Inspection Guide
- Storage Statement: Store in dry location
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service