Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MF-Millipore Membrane
Products
Product Category
membranes (13)
Brand
Millipore (13)
Diameter (mm)
Quality Segment
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
HAWP29325
MF-Millipore Membrane,
pore size 0.45 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
VSWP29325
MF-Millipore Membrane,
pore size 0.025 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
GSWP29325
MF-Millipore Membrane,
pore size 0.22 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
RAWP29325
MF-Millipore Membrane,
Expand
View Pricing
GSTF29325
MF-Millipore Membrane,
pore size 0.22 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
DAWP29325
MF-Millipore Membrane,
pore size 0.65 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
AAWP29325
MF-Millipore Membrane,
pore size 0.8 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
VCWP29325
MF-Millipore Membrane,
pore size 0.1 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
SCWP29325
MF-Millipore Membrane,
pore size 8.0 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
SMWP29325
MF-Millipore Membrane,
pore size 5.0 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
SSWP29325
MF-Millipore Membrane,
pore size 3.0 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
PHWP29325
MF-Millipore Membrane,
pore size 0.3 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing
RATF29325
MF-Millipore Membrane,
pore size 1.2 μm, filter diam. 293 mm, hydrophilic
Expand
View Pricing