Skip to Content
Merck
All Photos(4)

Key Documents

ABS1600

Sigma-Aldrich

Anti-phospho-UVRAG (Ser498)

from rabbit

Synonym(s):

UV radiation resistance-associated gene protein, p63

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
3

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer498)

Gene Information

human ... UVRAG(7405)

General description

UV radiation resistance-associated gene protein (UniProt: Q9P2Y5; also known as p63) is encoded by the UVRAG gene (Gene ID: 7405) in human. UVRAG is a key autophagic tumor suppressor that is mutated in several common human cancers. It contains four functional domains, a proline-rich domain, a lipid-binding C2 domain, a Beclin1-binding coiled-coil domain (CCD) and a C-terminal domain. It is a critical regulator of intracellular membrane trafficking, autophagy, and chromosomal stability. UVRAG is also involved in regulation of the COPI-dependent retrograde transport from Golgi and the endoplasmic reticulum by associating with the NRZ complex. Its function is dependent on its binding to phosphatidylinositol 3-phosphate (PtdIns3P). During autophagy UVRAG acts as regulatory subunit of the alternative PI3K complex II (PI3KC3-C2) that mediates formation of phosphatidylinositol 3-phosphate and is believed to be involved in maturation of autophagosomes and endocytosis. It is involved in maturation of autophagosomes and degradative endocytic trafficking independently of Beclin-1, but depends on its association with a class C Vps complex. UVRAG is also shown to enhance class C Vps complex association with a SNARE complex and promote fusogenic SNARE complex formation during late endocytic membrane fusion. UVRAG is also reported to promote DNA double-strand break repair by association with DNA-dependent protein kinase complex DNA-PK and activating it in non-homologous end joining. Disruption of UVRAG is shown to increase genetic instability and sensitivity of cells to irradiation. (Ref: Zhao, Z et al. (2012). Dev. Cell 22(5): 1001-1016).

Specificity

This polyclonal antibody detects human UVRAG phosphorylated on Serine 498.

Immunogen

A linear peptide corresponding to 15 amino acids from the C-terminal half of human UVRAG surrounding phosphoserine 498.

Application

Detect UV radiation resistance-associated gene protein using this rabbit polyclonal Anti-phospho-UVRAG (Ser498), Cat. No. ABS1600 that has been tested in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected phospho-UVRAG (Ser498) in TSC1 KO MEFs and treated with Torin1 (Kim, Y.M., et. al. (2015). Mol Cell. 57(2):207-18).

Western Blotting Analysis: A representative lot detected phospho-UVRAG (Ser498) in HA-UVRAG+myc-Rheb2+DMSO. (Courtesy of Dr. Do-Hyung Kim at University of Minnesota).


Quality

Evaluated by Western Blotting in HEK293 cells transfected with HA-UVRAG+myc-Rheb2+DMSO (vehicle).

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-UVRAG (Ser498) in HEK293 cells transfected with HA-UVRAG+myc-Rheb2+DMSO (vehicle).

Target description

~98 kDa observed; 78.15 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, pH7.4 with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Yuan et al.
International journal of molecular medicine, 42(6), 3562-3570 (2018-09-18)
Ultraviolet radiation resistance‑associated gene (UVRAG) regulates autophagy by promoting the formation and maturation of autophagosomes. The aim of the present study was to investigate the effects of UVRAG and UVRAG‑targeting miRNA on the regulation of autophagy and apoptosis in gastric
Toshiharu Fujita et al.
Journal of cell science, 134(6) (2021-02-17)
TFEB, a basic helix-loop-helix transcription factor, is a master regulator of autophagy, lysosome biogenesis and lipid catabolism. Compared to posttranslational regulation of TFEB, the regulation of TFEB mRNA stability remains relatively uncharacterized. In this study, we identified the mRNA-binding protein
Christine Quach et al.
Nature communications, 10(1), 5681-5681 (2019-12-14)
Aberrant autophagy is a major risk factor for inflammatory diseases and cancer. However, the genetic basis and underlying mechanisms are less established. UVRAG is a tumor suppressor candidate involved in autophagy, which is truncated in cancers by a frameshift (FS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service