Skip to Content
Merck
All Photos(2)

Key Documents

ABN2268M

Sigma-Aldrich

Anti-proSAAS

serum, from rabbit

Synonym(s):

IA-4, Proprotein convertase subtilisin/kexin type 1 inhibitor, Proprotein convertase 1 inhibitor, pro-SAAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IF
WB
species reactivity:
rat, mouse, human
technique(s):
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PCSK1N(27344)
mouse ... Pcsk1N(30052)

General description

ProSAAS (UniProt: Q9QXV0; also known as IA-4, Proprotein convertase subtilisin/kexin type 1 inhibitor, Proprotein convertase 1 inhibitor, pro-SAAS) is encoded by the Pcsk1n gene (Gene ID: 30052) in murine species. ProSAAS is a small secretory protein that is primarily expressed in neural and neuroendocrine tissues and acts as an anti-aggregant chaperone. Its presence appears to be restricted only to vertebrates. Mammalian proSAAS does not undergo either glycosylation or phosphorylation, but undergoes proteolytic cleavage at several pairs of basic residues that leaves its core region intact. ProSAAS is known to interact with proprotein convertase subtilisin/kexin type 1 (PCSK1) via the C-terminal inhibitory domain (aa 219-258). In the neural tissues proSAAS exhibits neuroprotective effects and is shown to block alpha-synuclein-induced cytotoxicity in cultures of dopaminergic neurons and in SH-SY5Y cells. In vitro studies have shown that proSAAS can also prevent the fibrillation of amyloid beta 1-42 peptide in a dose-dependent manner and residues 97-180 are sufficient for its anti-aggregation function against amyloid peptides. Inclusion of recombinant proSAAS in the medium of Neuro2a cells is shown to block the neurocytotoxic effect of amyloid peptide 1-42.

Specificity

This rabbit polyclonal antibody detects proSAAS in human, mouse, and rat cells. It targets an epitope within the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to first 180 amino acids from murine ProSAAS.

Application

Anti-proSAAS Antibody, Cat. No. ABN2268, is a highly specific rabbit polyclonal antibody that targets ProSAAS and has been tested in Immunocytochemistry, Immunofluorescence, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected proSAAS in AtT-20 cells. (Courtesy of Dr. Iris Lindberg @ University of Maryland).

Immunofluorescence Analysis: A representative lot detected proSAAS in Immunofluorescence applications (Jarvela, T.S., et. al. (2016). Proc Natl Acad Sci USA. 113(32):E4708-15; Hoshino, A., et. al. (2014). J Neurochem. 128(3):419-30; Guest, P.C., et. al. (2002). J Endocrinol. 175(3):637-47).

Western Blotting Analysis: A representative lot detected proSAAS in A immunoprecipitated from mouse brain lysates (Hoshino, A., et. al. (2014). J Neurochem. 128(3):419-30). Notes: Treating blots with 1% glutaraldehyde in PBS for 30 min prior to blocking significantly increased the proSAAS signal and reduces the background.

Immunocytochemistry Analysis: A representative lot detected proSAAS in SH-SY5Y cells (Jarvela, T.S., et. al. (2016). Proc Natl Acad Sci USA. 113(32):E4708-15).

Quality

Evaluated by Western Blotting in His-tagged mouse proSAAS recombinant protein.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected His-tagged mouse proSAAS recombinant protein. Note: For most biological samples a range of 1:1000 for Western blotting and 1:500 for IHC is recommended.

Target description

~19 kDa observed; 27.27 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service