Skip to Content
Merck
All Photos(1)

Key Documents

AB3607

Sigma-Aldrich

Anti-ASC Antibody

Chemicon®, from rabbit

Synonym(s):

TMS1, CARD5, Apoptosis-associated Speck-like Protein containing a CARD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
7

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD) containing molecules and a caspase family of proteases. CARD containing cell death regulators include RAIDD, RICK, Bcl10, Apaf-1, ARC, caspase-2 and caspase-9. A novel CARD domain containing protein has been identified in human and mouse and designated ASC and TMS1. Ectopic expression of ASC/TMS1 induced apoptosis through activation of caspase-9 and inhibited the survival of human breast cancer cells. Overexpression of ASC/TMS1 induced DNA fragmentation. ASC/TMS1 is expressed in a variety of human and mouse tissues.

Specificity

Detects a 25 kDa band by western blot, corresponding to ASC/TMS1.

Immunogen

Peptide corresponding to aa 182-195 (RESQSYLVEDLERS) of human ASC.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-ASC Antibody is validated for use in WB for the detection of ASC.
Western blot: 1:500

HL60 whole cell lysate can be used as a positive control.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Format: Purified

Storage and Stability

Maintain at 4°C for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Zhao et al.
Journal of Crohn's & colitis, 15(4), 647-664 (2020-10-07)
Crohn's disease [CD] is a chronic, relapsing and incurable inflammatory disorder. Micro RNAs [miRNAs], which modulate gene expression by binding to mRNAs, may make significant contributions to understanding the complex pathobiology and aetiology of CD. This study aimed to investigate
Y Xing et al.
Cell death & disease, 7(8), e2322-e2322 (2016-08-05)
Autophagosomes derived from tumor cells, also referred to as defective ribosomal products in blebs (DRibbles), have been previously shown to stimulate potent T-cell responses and mediate tumor regression when used as therapeutic cancer vaccines in multiple preclinical cancer models. In
Rekha R Garg et al.
The Journal of biological chemistry, 294(21), 8480-8489 (2019-04-04)
Among the repertoire of immunoregulatory proteins encoded by myxoma virus, M013 is a viral homologue of the viral pyrin domain-only protein (vPOP) family. In myeloid cells, M013 protein has been shown to inhibit both the inflammasome and NF-κB signaling pathways
Yunchao Shi et al.
Brain and behavior, 13(5), e2980-e2980 (2023-03-30)
Cognitive impairment is a critical complication of acute respiratory distress syndrome (ARDS). However, effective interventions are lacking. Growing evidence demonstrates that c-Jun N-terminal kinase (JNK)-mediated neuroinflammation is involved in the development of ARDS. Therefore, we hypothesized that the JNK pathway
Shuyun Yang et al.
Evidence-based complementary and alternative medicine : eCAM, 2021, 1821220-1821220 (2021-10-05)
Acne vulgaris (AV) is a chronic skin disease involving inflammation of the pilosebaceous units. Propionibacterium acnes (P. acnes) hypercolonization is one pathogenic factor for AV. P. acnes that triggers interleukin-1β (IL-1β) by activating the pyrin domain-containing 3 protein (NLRP3) inflammasome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service