Skip to Content
MilliporeSigma

Skip To

E6909

ECM Gel from Engelbreth-Holm-Swarm murine sarcoma

growth-factor reduced, without phenol red, liquid, BioReagent, suitable for cell culture

Synonym(s):

GFR Basement Membrane Extract (BME)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mL
Check Cart for Availability
$424.00
10 mL
Check Cart for Availability
$726.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:

$424.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

BioReagent

form

liquid

concentration

7-9 mg/mL

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Application

ECM gel can be used with epithelial cells, endothelial cells, muscle cells, nerve cells and tumor cells.
ECM Gel from Engelbreth-Holm-Swarm murine sarcoma has been used to coat tissue culture plates for different glioma subtypes and C2C12 muscle cells. It has also been used as a scaffold to increase myotube attachment, maturation and survival.

Biochem/physiol Actions

The ECM gel will undergo thermally activated polymerization when brought to 20-40°C, forming a reconstituted basement membrane. PC12 cells will show neurite formation within 2 days when they are grown on a thin layer of this ECM gel.

Every mouse colony used for the production of this product is routinely screened for several pathogens. Tested and found negative for LDEV

Preparation Note

This product is prepared from Engelbreth-Holm-Swarm sarcoma in mice; dialyzed against chloroform. Gel should be thawed overnight at 2-8°C before use and dispensed to a multiwall plate using a plate and pipettes that are pre-cooled to 2-8°C. The gel may contain precipitates which do not influence its activity. The gel may also be diluted up to twofold with 2-8°C Dulbecco′s Modified Eagle′s Medium, and should be done before gel is added to the plate. The product will gel within 5 minutes at 20°C. For prolonged manipulations, work should be conducted below 10°C. Cells can be plated on top of a thin gel layer of 0.5 mm or cultured inside a 1 mm layer. If you culture cells inside, add the cells to the gel prior to plating at a recommended density of 3-4 x 104 cells per mL. To dissociate cells from this gel, use protease in PBS without calcium, magnesium, or EDTA, at a concentration of 0.6-2.4 units per mL.

Other Notes

ECM was prepared to a protein concentration of 8-12 mg/mL, containing laminin as a major component, collagen type IV, heparin sulfate proteoglycan, entactin, and other minor components. Addition of collagen type IV to the ECM gel increases polymerization.
The growth factor reduction modification allows for more control of medium, better interpretation of results and increased polymerization. The medium also lacks phenol red, which can interfere with some analyses.

Disclaimer

For long term storage, keep product at -20°C. The ECM gel may be stored at 2-8°C for up to 72 hours.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E1270B1522D1145
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

liquid

form

liquid

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

shipped in

ambient

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

300

Quality Level

400

Quality Level

400

product line

BioReagent

product line

BioReagent

product line

-

product line

-


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–8 of 8 Questions  
  1. ECM cat:E6909 I would like to know if this product could be used in vivo to inject tumor cells into mouse liver.Its description says reduced growth factor but I would like to know if it contains anything, even in a low %,that can induce cell proliferation

    1 answer
    1. This product is not tested for in vivo use. The Quality testing does include neurite formation in cell culture. The end user would have to determine suitability for use in animal models. This ECM product undergoes an additional purification step which depleted growth factors. However, the material is not tested for the presence of growth factors post-purification. See the links below to review additional information regarding the presence and depletion of growth factors in Extracellular Matrix from mouse sarcoma:
      https://www.researchgate.net/publication/41454358_Matrigel_A_complex_protein_mixture_required_for_optimal_growth_of_cell_culture
      https://www.pnas.org/doi/epdf/10.1073/pnas.87.10.4002
      https://www.sciencedirect.com/science/article/pii/S1044579X05000313#bib16
      https://www.sciencedirect.com/science/article/abs/pii/001448279290397Q

      Helpful?

  2. What is the application of ECM for tube assay and the impact of reduced growth factors?

    1 answer
    1. I recommend the following publication for a comparison study on ECM vs ECM with reduced growth factor: J Vis Exp. 2016 Aug 14;(114):54074. doi: 10.3791/54074. Strategic Endothelial Cell Tube Formation Assay: Comparing Extracellular Matrix and Growth Factor Reduced Extracellular Matrix. PMID: 27585062.

      J Vis Exp. 2016 Aug 14;(114):54074. doi: 10.3791/54074. Strategic Endothelial Cell Tube Formation Assay: Comparing Extracellular Matrix and Growth Factor Reduced Extracellular Matrix. PMID: 27585062.

      Helpful?

  3. Is it possible to dissociate cells using Accutase when they are attached to the ECM gel E6909?

    1 answer
    1. Yes, it is possible to dissociate cells using Accutase when they are attached to the ECM gel E6909. It is advisable to start with a small-scale trial to assess its suitability.

      Helpful?

  4. Do Products E1270 and E6909, ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, contain phenol red?

    1 answer
    1. Product E1270 ECM gel does contain phenol-red from the Dulbecco's modified Eagle's medium component of the product.  Product No. E6909, ECM gel, growth factor reduced, is phenol red free.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Are Products E1270 and E6909, ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, growth factor reduced like Matrigel (from BD)?

    1 answer
    1. Product No. E1270 is not growth factor reduced. Product No. E6909 is ECM gel, growth factor reduced.

      Helpful?

  7. Are ECM Gels, Products E1270 and E6909, the same as Matrigel (from BD)?

    1 answer
    1. Product No. E1270, ECM gel is the equivalent of Matrigel from BD only produced by Sigma-Aldrich.

      Helpful?

  8. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

Reviews

No rating value

Active Filters