Skip to Content
Merck
All Photos(1)

Key Documents

AB1538

Sigma-Aldrich

Anti-Dopamine β Hydroxylase Antibody, NT

Chemicon®, from rabbit

Synonym(s):

DBH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, bovine, sheep

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DBH(1621)

Specificity

Dopamine Beta Hydroxylase (DbetaH) N-terminal. By immunoblot the antibody specifically reacts with a single (or double) band(s) of M.W. »70-75,000 in samples of SDS-solubilized, DTT-reduced human adrenal medulla subjected to SDS-PAGE.

Immunogen

Epitope: N-terminus
Synthetic peptide, CSAPRESPLPYHIPLDPEG-amide, cloned from human (DbetaH) cDNA (Nagatsu et al. 1990).

Application

Detect Dopamine β Hydroxylase using this Anti-Dopamine β Hydroxylase Antibody, N-terminus validated for use in IH & WB.
Immunoblotting: 1:250-1:1,000.

Immunohistochemistry: 1:250-1:1,000, staining was evaluated in the locus coeruleus of formaldehyde-fixed, cryopreserved human and monkey brain Showed weak staining on both human and monkey.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Physical form

Liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20 to -70°C in undiluted aliquots for up to six months. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Josée Wallman et al.
The European journal of neuroscience, 33(8), 1519-1532 (2011-03-08)
This study aimed to provide a first detailed description of the serotonin (5-hydroxytryptamine, 5-HT) innervation of the human basal ganglia under nonpathological conditions. We applied an immunohistochemical approach to postmortem human brain material with antibodies directed against the 5-HT transporter
M E Da Vitoria Lobo et al.
Frontiers in pain research (Lausanne, Switzerland), 4, 1190440-1190440 (2023-06-16)
Chronic pain is a prevalent physically debilitating health-related morbidity. Frontline analgesics are inadequate, providing only partial pain relief in only a proportion of the patient cohort. Here, we explore whether alterations in spinal cord vascular perfusion are a factor in
Keita Harada et al.
Journal of neurochemistry, 114(2), 617-626 (2010-05-19)
Neurons and certain kinds of endocrine cells, such as adrenal medullary (AM) cells, have large dense-core vesicles (LDCVs) and synaptic vesicles or synaptic-like microvesicles (SLMVs). These secretory vesicles differ in Ca(2+) sensitivity and contain different signaling substances. We have recently
Georg Gussak et al.
JCI insight, 4(20) (2019-09-11)
Atrial fibrillation (AF) is the most common heart rhythm disorder and a major cause of stroke. Unfortunately, current therapies for AF are suboptimal, largely because the molecular mechanisms underlying AF are poorly understood. Since the autonomic nervous system is thought
Günter U Höglinger et al.
The Journal of comparative neurology, 522(10), 2336-2348 (2014-01-16)
Adult neurogenesis, a specific form of brain plasticity in mammals occurring in the subventricular zone (SVZ) and the subgranular zone (SGZ), is subject to complex regulations. Dopamine has been identified as a regulatory factor in rodents, nonhuman primates, and humans.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service