Skip to Content
Merck
All Photos(1)

Key Documents

8.20866

Sigma-Aldrich

1-Naphthylammonium chloride

for synthesis

Synonym(s):

1-Naphthylammonium chloride, 1-Naphthylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10ClN
CAS Number:
Molecular Weight:
179.65
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-014-3
NACRES:
NA.22
form:
powder
Assay:
≥98.0% (HPLC)

Quality Level

Assay

≥98.0% (HPLC)

form

powder

potency

680 mg/kg LD50, oral (Rat)

mp

272-275 °C

solubility

37.7 g/L

storage temp.

2-30°C

SMILES string

[Cl-].[N+H3]c1c2c(ccc1)cccc2

InChI

1S/C10H9N.ClH/c11-10-7-3-5-8-4-1-2-6-9(8)10;/h1-7H,11H2;1H

InChI key

FOKKJVHTXPJHEN-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service