Skip to Content
Merck
All Photos(1)

Key Documents

8.20639

Sigma-Aldrich

1-Hexene

Technipur®, for synthesis

Synonym(s):

Butylethylene, Hex-1-ene, Hexylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1209240
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21
grade:
for synthesis
bp:
60-66 °C (lit.)
vapor pressure:
155 mmHg ( 21.1 °C)
412 hPa ( 38 °C)

grade

for synthesis

Quality Level

vapor density

3 (vs air)

vapor pressure

155 mmHg ( 21.1 °C)
412 hPa ( 38 °C)

autoignition temp.

487 °F

refractive index

n20/D 1.388 (lit.)

bp

60-66 °C (lit.)

mp

-140 °C

density

0.678 g/mL at 25 °C (lit.)

SMILES string

CCCCC=C

InChI

1S/C6H12/c1-3-5-6-4-2/h3H,1,4-6H2,2H3

InChI key

LIKMAJRDDDTEIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexene is a linear alpha-olefin, which carries a double bond at the alpha position. It is a widely used raw material in the production of synthetic rubbers, elastomers, plasticizers, mercaptans, fatty acids, and synthetic resins. 1-Hexene is also used as a solvent in paints, printing inks, resins, and waxes.

Application

1-Hexene can be used as:
  • A monomer in the production of polyhexene-1 (PHe) elastomer material via metal-catalyzed homopolymerization reaction. This elastomeric material can be further utilized in the modification of plastic and rubber products.
  • A comonomer in the manufacture of polyethylenes such as linear low-density polyethylene (LDPE) and high-density polyethylene (HDPE). These materials are widely used in everyday products such as plastic bags, packaging films, agricultural films, and containers.
  • A solvent in the separation of olefins and paraffins.

It can also be used in copolymerization reactions along with 1-butylene or propylene to synthesize thermoplastic copolymers to increase the impact strength, elongation at break, density, and melt flow index and to impart good stiffness-flexibility balance.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on Properties of PP Modified by Butylene-Hexene Copolymer
Z Yong-xian, et al.
China plastics, 3 (2006)
Excess molar enthalpy measurement and modelling of (oxygenated compounds+ hydrocarbon) mixtures: Binary and ternary mixtures containing 2-(2-methoxyethoxy) ethanol, 1-hexene and cyclohexane at (298.15 and 313.15) K
Lifi M, et al.
The Journal of Chemical Thermodynamics, 149, 106171-106171 (2020)
Liquid-liquid equilibria of ternary systems of 1-hexene/hexane and extraction solvent
Zhu L, et al.
Chemical Papers, 70(5), 585-593 (2016)
Study on the synthesis of hexene-1 catalyzed by Ziegler-Natta catalyst and polyhexene-1 applications
Yan Y, et al.
EPL, 19(1), 511-518 (2019)
Copolymerization of propylene with higher a-olefins by a Pyridylamidohafnium catalyst: An effective approach to polypropylene-based elastomer
Yang F, et al.
Polymers, 12(1), 89-89 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service