Skip to Content
Merck
All Photos(1)

Key Documents

8.20526

Sigma-Aldrich

meso-1,2-Diphenyl-1,2-ethanediol

for synthesis

Synonym(s):

meso-1,2-Diphenyl-1,2-ethanediol, meso-Hydrobenzoin, meso-1,2-Diphenylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH(C6H5)CH(C6H5)OH
CAS Number:
Molecular Weight:
214.26
MDL number:
UNSPSC Code:
12352104
EC Index Number:
207-758-3
NACRES:
NA.22

Quality Level

Assay

≥96.0% (HPLC)

form

powder

mp

139 °C

solubility

slightly soluble 2.5 g/L

storage temp.

2-30°C

SMILES string

OC(C(O)c2ccccc2)c1ccccc1

InChI

1S/C14H14O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-16H

InChI key

IHPDTPWNFBQHEB-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 133 °C
Melting range (upper value): ≤ 137 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service