Skip to Content
Merck
All Photos(1)

Documents

8.14068

Sigma-Aldrich

Tetrapropylammonium hydroxide

(40% solution in water) for synthesis

Synonym(s):

N,N,N-tripropyl-1-propanaminium hydroxide, Tetra-n-propylammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.22

Quality Level

form

liquid

pH

>7 (20 °C in H2O, undiluted)

bp

102 °C/1013 hPa

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

General description

Tetrapropylammonium hydroxide (TPAOH) is a simple, affordable, and readily available quaternary ammonium base mainly utilized as a structure-directing agent for producing zeolites. It is found to be a good precursor to synthesis nanotubes. TPAOH is used as surface-active agent, phase transfer catalyst in organic synthesis.

Application

Tetrapropylammonium hydroxide is used as:
  • Structure directing agent (SDA) in the synthesis of zeolite socony mobil-5 (Zeolite ZSM-5).
  • A phase transfer catalyst in the methylation of phenoxy acids in a superheated water extract from sand and soil samples.

Analysis Note

Assay (acidimetric): 38.0 - 42.0 %
Density (d 20 °C/ 4 °C): 0.998 - 1.010

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbonization mechanism of tetrapropylammonium-hydroxide in channels of AlPO4-5 single crystals
Zhai JP, et al.
Chemistry of Materials, 18(6), 1505-1511 (2006)
Surface-active hollow titanosilicate particles as a Pickering interfacial catalyst for liquid-phase alkene epoxidation reactions
Guojun LV, et al.
Langmuir, 34(1), 302-310 (2018)
Synthesis of ZSM- 5 zeolite using Bayat natural zeolite as silica and alumina source
Rohayati, Krisnandi YK and Sihombing R
AIP Conference Proceedings, 1862(1), 030094-030094 (2017)
Orapin Chienthavorn et al.
Journal of chromatography. A, 1152(1-2), 268-273 (2007-01-11)
Phase transfer catalytic methylation was applied to directly derivatise chlorophenoxy acid herbicides in superheated water extracts from sand and soil samples. The extractions were carried out at 120 degrees C statically for 5 min and then dynamically for 10 min

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service