Skip to Content
Merck
All Photos(1)

Key Documents

86578

Sigma-Aldrich

Tetraethyl orthosilicate

≥99.0% (GC)

Synonym(s):

Orthosilicic acid tetraethyl ester, Silicon tetraethoxide, TEOS, Tetraethoxysilane, Tetraethoxysilicon(IV), Tetraethyl silicate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(OC2H5)4
CAS Number:
Molecular Weight:
208.33
Beilstein:
1422225
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
technique(s):
GC/GC: suitable
bp:
168 °C (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

vapor density

7.2 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

technique(s)

GC/GC: suitable

refractive index

n20/D 1.382 (lit.)
n20/D 1.383

bp

168 °C (lit.)

density

0.933 g/mL at 20 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)OCC

InChI

1S/C8H20O4Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-8H2,1-4H3

InChI key

BOTDANWDWHJENH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetraethyl orthosilicate (TEOS) is an inorganic material that can be used as a silica source for the synthesis of silica-based materials such as silicon dioxide, silicon oxycarbides, silanol, siloxane polymer, and organosilicon thin films for a variety of applications. It can also be used in the synthesis of blended membranes, and the production of aerogel. Other applications include coatings for carpets and other objects.

Application

Commonly used as a precursor to prepare xerogel
Tetraethyl orthosilicate (TEOS) can be used:
  • As a reagent along with ferric chloride in the synthesis of dihydropyrimidinones.
  • As a modifier in fabrication of humidity sensing material, poly(2-acrylamido-2-methylpropane sulfonate) (poly-AMPS).
  • As a medium to synthesize triarylamines and diaryl ethers via copper catalyzed, ligand free-Ullmann reaction.
  • As a precursor for the preparation of silica xerogel which finds application in pharmaceutical drug carriers.
  • As a precursor for the synthesis of spherical silica particles by Stober process.

Will interact with dodecylamine in the formation of intercalation compounds of H+-magadiite and used in a study of mixed-metal bioactive glasses.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of synthetic carpet using chitosan-titania nanocomposite for self-cleaning purposes
Sulaeman MI, et al.
AIP Conference Proceedings, 2175(1), 020077-020077 (2019)
Use of poly (2-acrylamido-2-methylpropane sulfonate) modified with tetraethyl orthosilicate as sensing material for measurement of humidity
Su PG, et al.
Analytica Chimica Acta, 1-2(449), 103-109 (2001)
Ferric chloride/tetraethyl orthosilicate as an efficient system for synthesis of dihydropyrimidinones by Biginelli reaction
Cepanec I, et al.
Tetrahedron, 17(61), 4275-4280 (2005)
The Organosilicon thin Film Deposited Using an Atmospheric Pressure Dual-Frequency 50 kHz/33 MHz Frequency MicroPlasma Jet
Yuan QH, et al.
Contributions to Plasma Physics, 56(9), 870-877 (2016)
Silica xerogels as pharmaceutical drug carriers
Quintanar-Guerrero D, et al.
Expert Opinion on Drug Delivery, 5(6), 485-498 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service