810243
Bovine Serum Albumin Sulfhydryl Modified
≥98%, USA origin, Tested For IgG, Protein standard: suitable, enzyme immunoassay: suitable
About This Item
Recommended Products
Product Name
Bovine Serum Albumin Sulfhydryl Modified, pack of 1 kg
grade
special quality (Specialty Grade)
Quality Level
Assay
≥98% (purity)
form
lyophilized powder
shelf life
48 mo.
mol wt
~66 kDa
purified by
heat shock fractionation
packaging
pack of 1 kg
origin
USA origin
Free Sulfhydrl Content content
0.0-1.0 mmol/mol
technique(s)
ELISA: suitable
enzyme immunoassay: suitable
protein quantification: suitable
impurities
Tested For IgG
loss
≤5%
pH
6.8-7.5
anion traces
chloride (Cl-): 0.0-6.0 mg/g
cation traces
Na: 0.0-15.0 mg/g
foreign activity
Bluetongue, none detected
Vesicular Stomatitis, none detected
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service