Skip to Content
Merck
All Photos(1)

Key Documents

8.06373

Sigma-Aldrich

Sodium borohydride

fine granular for synthesis

Synonym(s):

Sodium borohydride, Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H4BNa
CAS Number:
Molecular Weight:
37.83
MDL number:
UNSPSC Code:
12352306
EC Index Number:
241-004-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 25 °C)

Quality Level

form

solid

autoignition temp.

220 °C

potency

160 mg/kg LD50, oral (Rat)
230 mg/kg LD50, skin (Rabbit)

expl. lim.

3.02 % (v/v)

reaction suitability

reagent type: reductant

pH

11 (20 °C, 10 g/L in H2O)

bp

500 °C (decomposition)

mp

>360 °C (slow decomposition)

transition temp

flash point 69 °C

density

1.07 g/cm3 at 20 °C

bulk density

350‑500 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[B-H2]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium borohydride (NaBH4) is the most commonly available borohydride used in several industrial applications. It is considered as a selective reducing agent due to its ability to selectively reduce aldehydes and ketones into their corresponding alcohols in the presence of other functional groups such as nitro, ester, amide, lactone, epoxide, and nitriles. Ethanol and methanol are usually employed as solvents in NaBH4 reduction reactions.[1]

Application

Sodium borohydride (fine granular) can be used as a reducing agent to prepare:
  • Zero valent iron nanoparticles supported on mesoporous carbon (Starbon) by in situ reductions of Fe3+ ions.[2]
  • Ag nanoparticles on graphene using AgNO3 as a precursor.[3]
  • Ag-Pd/ZrO2 catalytic system applicable in plasmonic catalysis.[4] It can also be used as a source to store as well as generate hydrogen for energy-carrier applications.[5]

Analysis Note

Assay (hypochlorite): ≥ 98.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sodium Borohydride
Banfi Luca, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2014)
Hydrogen production from solid sodium borohydride
Gislon P, et al.
International Journal of Hydrogen Energy, 34(2), 929-937 (2009)
Synthesis and characterization of robust zero valent iron/mesoporous carbon composites and their applications in arsenic removal
Baikousi M, et al.
Carbon, 93, 636-647 (2015)
Erandi Peiris et al.
Journal of visualized experiments : JoVE, (162) (2020-09-08)
Localized surface plasmon resonance (LSPR) in plasmonic nanoparticles (NPs) can accelerate and control the selectivity of a variety of molecular transformations. This opens possibilities for the use of visible or near-IR light as a sustainable input to drive and control
Ag-stabilized few-layer graphene dispersions in low boiling point solvents for versatile nonlinear optical applications
Sun Zhenyu, et al.
Carbon, 62, 182-192 (2013)

Protocols

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service