Skip to Content
Merck
All Photos(1)

Key Documents

8.01336

Sigma-Aldrich

Pivalic acid

for synthesis

Synonym(s):

Pivalic acid, Neopentanoic acid, 2,2-Dimethylpropionic acid, Trimethylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOOH
CAS Number:
Molecular Weight:
102.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-922-5
NACRES:
NA.22

vapor pressure

0.85 hPa ( 20 °C)

Quality Level

form

crystals

potency

900 mg/kg LD50, oral (Rat)
1900 mg/kg LD50, skin (Rat)

expl. lim.

1.6 % (v/v)

mp

32-35 °C

transition temp

flash point 64 °C

solubility

25 g/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)C(C)(C)C

InChI

1S/C5H10O2/c1-5(2,3)4(6)7/h1-3H3,(H,6,7)

InChI key

IUGYQRQAERSCNH-UHFFFAOYSA-N

Application


  • Regiodivergent synthesis involving pivalic acid: A ligand-controlled regiodivergent C-H alkenylation process using pivalic acid, advancing the synthesis of complex indazoles and potentially impacting pharmaceutical compound development (Kim et al., 2017).

  • Pivalic acid in radiochemical syntheses: The study utilized pivalic acid derivatives for the synthesis of novel radioiodinated pharmaceuticals for hypoxia imaging, highlighting its utility in diagnostic and therapeutic applications in the medical field (Elsaidi et al., 2019).

  • Capillary electrophoretic applications of pivalic acid: Research explored new methods of cationic electrophoretic focusing using pivalic acid for high-sensitivity analysis of water contaminants, providing insights into environmental monitoring techniques (Malá and Gebauer, 2022).

  • Cyclative coupling using pivalic acid: Zhuang et al. described a novel cyclative C-H/C-H coupling technique facilitated by pivalic acid, which rapidly constructs complex molecular frameworks like tetralin and chromane, useful in drug discovery and development (Zhuang et al., 2021).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service