Skip to Content
Merck
All Photos(2)

Key Documents

240370

Sigma-Aldrich

Valeric acid

≥99%

Synonym(s):

n-Valeric acid, Pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COOH
CAS Number:
Molecular Weight:
102.13
Beilstein:
969454
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

707 °F

expl. lim.

7.6 %

refractive index

n20/D 1.408 (lit.)

bp

110-111 °C/10 mmHg (lit.)
185 °C (lit.)

mp

−20-−18 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCC(O)=O

InChI

1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)

InChI key

NQPDZGIKBAWPEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Valeric acid (VA) also known as pentanoic acid, is a short-chain fatty acid used to synthesize valeric biofuels.[1]

Application

Sex attractant of the sugar beet wireworm, Limonius californicus.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeric acid lowers arterial blood pressure in rats
Onyszkiewicz M, et al.
European Journal of Pharmacology, 877, 173086-173086 (2020)
Chun-Chieh Lin et al.
eLife, 4 (2015-10-01)
Animals use olfactory cues for navigating complex environments. Food odors in particular provide crucial information regarding potential foraging sites. Many behaviors occur at food sites, yet how food odors regulate such behaviors at these sites is unclear. Using Drosophila melanogaster
M F Böttcher et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 30(11), 1590-1596 (2000-11-09)
The prevalence of allergic diseases has increased particularly over the past 30-40 years. A reduced microbial stimulation during infancy may result in a development of a disturbed balance between Th1- and Th2-like immunity. The gut flora is, quantitatively, the most
D M Saunders et al.
The Australian & New Zealand journal of obstetrics & gynaecology, 18(3), 198-201 (1978-08-01)
The effect of oestradiol valerate on levels of blood cholesterol, triglycerides, free oestradiol and coagulation factors, as well as on urinary oestrogens and free cortisol was measured in 10 oöphorectomised women. Despite the very high urinary oestrogen levles obtained, there
Valeric biofuels: a platform of cellulosic transportation fuels.
Jean-Paul Lange et al.
Angewandte Chemie (International ed. in English), 49(26), 4479-4483 (2010-05-07)

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service