Skip to Content
Merck
All Photos(1)

Key Documents

688000

Sigma-Aldrich

Y-27632

≥95% (HPLC), solid, Rho kinase inhibitor, Calbiochem

Synonym(s):

Y-27632, (R)-(+)-trans-N-(4-Pyridyl)-4-(1-aminoethyl)-cyclohexanecarboxamide, 2HCl, ROCK Inhibitor, Rho Kinase Inhibitor VI, ROCK Inhibitor, (R)-(+)-trans-N-(4-Pyridyl)-4-(1-aminoethyl)-cyclohexanecarboxamide, 2HCl, Rho Kinase Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O · 2HCl · xH2O
CAS Number:
Molecular Weight:
320.26 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N[C@@H](C1CCC(CC1)C(=O)Nc2ccncc2)C

InChI

1S/C14H21N3O/c1-10(15)11-2-4-12(5-3-11)14(18)17-13-6-8-16-9-7-13/h6-12H,2-5,15H2,1H3,(H,16,17,18)/t10-,11?,12?/m1/s1

InChI key

IYOZTVGMEWJPKR-VOMCLLRMSA-N

General description

A highly potent, cell-permeable, reversible, and selective inhibitor of Rho-associated protein kinases (Ki = 140 nM for p160ROCK). Also inhibits ROCK-II with equal potency. The inhibition is competitive with respect to ATP. Exhibits 10- to 50-fold lower affinity for PKCε than p160ROCK. Does not affect the activity of p21-activated protein kinase (PAK) even at higher concentrations (~100 µM). Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contraction. Prevents apoptosis and enhances the survival and cloning efficiency of dissociated hES cells without affecting their pluripotency. Pretreatment with Y-27632 reduces the risk of lung injury in rat models of sepsis. A 5 mM (500 µg/296 µl) solution of Y-27632 (Cat. No. 688001) in H2O and a 10 mM solution in DMSO (Cat. No. 688002) are also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p160 Rho-associated protein kinases (ROCK)
Product competes with ATP.
Reversible: yes
Target Ki: 140 nM for p160 Rho-associated protein kinases (ROCK)

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 6 months at -20°C.

Other Notes

Cinel, I., et al. 2012. J. Thorac . Dis.4, 30.
Watanabe, K., et al. 2007. Nature Biotech.25, 681.
Chitaley, K., et al. 2001. Nat. Med.7, 119.
Davies, S.P., et al. 2000. Biochem. J. 351, 95.
Narumiya, S., et al. 2000. Methods Enzymol.325, 273.
Hirose, M., et al. 1998. J. Cell Biol.141, 1625.
Maekawa, M., et al. 1999. Science285, 895.
Uehata, M., et al. 1997. Nature389, 990.

Legal Information

Sold under license of PCT Application W098/06,433A1 and under license from Mitsubishi Pharma Tanabe Corporation.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica L Teo et al.
Molecular biology of the cell, 31(23), 2557-2569 (2020-09-10)
Epithelia can eliminate apoptotic cells by apical extrusion. This is a complex morphogenetic event where expulsion of the apoptotic cell is accompanied by rearrangement of its immediate neighbors to form a rosette. A key mechanism for extrusion is constriction of
Adrian Danescu et al.
Development (Cambridge, England), 148(9) (2021-03-25)
In the face, symmetry is established when bilateral streams of neural crest cells leave the neural tube at the same time, follow identical migration routes and then give rise to the facial prominences. However, developmental instability exists, particularly surrounding the
Ge Guo et al.
Cell stem cell, 28(6), 1040-1056 (2021-04-09)
Classic embryological experiments have established that the early mouse embryo develops via sequential lineage bifurcations. The first segregated lineage is the trophectoderm, essential for blastocyst formation. Mouse naive epiblast and derivative embryonic stem cells are restricted accordingly from producing trophectoderm.
Benoit Dehapiot et al.
PLoS biology, 19(9), e3001376-e3001376 (2021-09-08)
Mammalian oocyte meiotic divisions are highly asymmetric and produce a large haploid gamete and 2 small polar bodies. This relies on the ability of the cell to break symmetry and position its spindle close to the cortex before anaphase occurs.
Masaya Miyazaki et al.
International journal of molecular medicine, 48(4) (2021-09-02)
The autophagy‑lysosome system allows cells to adapt to environmental changes by regulating the degradation and recycling of cellular components, and to maintain homeostasis by removing aggregated proteins and defective organelles. Cyclin G‑associated kinase (GAK) is involved in the regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service