Skip to Content
Merck

SCM075

ROCK Inhibitor (Y-27632)

The ROCK Inhibitor (Y-27632) is available in a 5 mg format & has been optimized & validated for cell culture & Neuroscience applications.

Synonym(s):

Y-27632

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

mol wt

320.26 g/mol

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

-10 to -25°C

General description

ROCK Inhibitor (Y-27632) is a selective, ATP-competitive inhibitor of Rho-associated protein kinase (ROCK) including p160Rock (Ki=140nM) and Rock-II. Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contraction. One-hour treatment with 10uM Y-27632 blocks apoptosis of dissociated cultured human embryonic stem cells (hESCs), increases survival and cloning efficiency of hESCs without affecting their pluripotency1. Recently, some reports showed that the application of Y-27632 and Accutase (Cat. No. SCR005), a formulated mixture of digestive enzymes, enhances the post-thaw survival rate of single hESCs using conventional slow freezing and rapid thawing methods

Preparation Note

Before Reconstitution: Maintain at -20ºC, protected from light for 2 years. After Reconstitution: 1 year at -20ºC, protected from light


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library