Skip to Content
Merck
All Photos(1)

Key Documents

524628

Millipore

Phosphatase Inhibitor Cocktail IV

liquid, for the inhibition of serine/threonine and alkaline phosphatases

Synonym(s):

Phosphatase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

form:
liquid
storage temp.:
−20°C
Pricing and availability is not currently available.

Product Name

Phosphatase Inhibitor Cocktail Set IV, Phosphatase inhibitor Cocktail Set IV is a cocktail of three phosphatase inhibitors for the inhibition of both serine/threonine and alkaline phosphatases.

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cocktail of three phosphatase inhibitors for the inhibition of both serine/threonine and alkaline phosphatases. Available as a 1 ml vial or as a set of five 1 ml vials. Each vial contains a 1 ml solution with the following components: 500 µM Cantharidin (Cat. No. 210155), 2.5 mM (-)-p-Bromotetramisole oxalate, and 1 µM Calyculin A, Discodermia calyx (Cat. No. 208851).
A cocktail of three phosphatase inhibitors specially formulated for inhibition of alkaline phosphatases as well as serine/threonine protein phosphatases such as PP1 and PP2A. May be used with cell lysates and tissue extracts, including extracts containing detergents. Supplied as 1 ml or a set of five 1 ml vials. Each vial contains 1 ml phosphatase inhibitor cocktail solubilized in 95% DMSO, consisting of the following components:

Biochem/physiol Actions

Cell permeable: no
Primary Target
Serine/threonine and alkaline phosphatases
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic (H)

Physical form

In 95% DMSO.

Reconstitution

For long-term storage, aliquot and freeze (-20°C). Dilute 1:50 just prior to use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jooyoung Lim et al.
International journal of stem cells, 17(3), 284-297 (2024-04-12)
Acetaldehyde dehydrogenase 2 (ALDH2) is the second enzyme involved in the breakdown of acetaldehyde into acetic acid during the process of alcohol metabolism. Roughly 40% of East Asians carry one or two ALDH2*2 alleles, and the presence of ALDH2 genetic
Stephane Angers
Methods in molecular biology (Clifton, N.J.), 468, 223-230 (2008-12-23)
Multiple screens performed in Drosophila, Caenorhabditis elegans, Xenopus, and zebrafish have identified dozens of proteins participating in Wnt signal transduction. Epistasis experiments, enhancer and suppressor screens, and protein-protein interaction techniques have also been efficient at finding new pathway members, connecting
Heeju Han et al.
Scientific reports, 12(1), 6416-6416 (2022-04-21)
Post-inflammatory hyperpigmentation is a skin discoloration process that occurs following an inflammatory response or wound. As the skin begins to heal, macrophages first exhibit a proinflammatory phenotype (M1) during the early stages of tissue repair and then transition to a
Chunwan Lu et al.
Oncogene, 41(18), 2651-2662 (2022-03-31)
The role of glucose-6-phosphate dehydrogenase (G6PD) in human cancer is incompletely understood. In a metabolite screening, we observed that inhibition of H3K9 methylation suppressed aerobic glycolysis and enhances the PPP in human mesothelioma cells. Genome-wide screening identified G6PD as an
Angela Ka Wai Lai et al.
Neural regeneration research, 17(7), 1566-1575 (2021-12-18)
Reperfusion therapy is the preferred treatment for ischemic stroke, but is hindered by its short treatment window, especially in patients with diabetes whose reperfusion after prolonged ischemia is often accompanied by exacerbated hemorrhage. The mechanisms underlying exacerbated hemorrhage are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service