Skip to Content
Merck

Skip To

487663

p-Nitrophenyl Phosphate

This p-Nitrophenyl Phosphate is validated for use in ELISA.

Synonym(s):

Phosphate Ester, p-Nitrophenyl Ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
41116158
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

clearslightly yellow tint

shipped in

ambient

storage temp.

2-8°C

General description

p-Nitrophenyl phosphate is the most common substrate for alkaline phosphatase-based ELISA assays. Hydrolysis of the phosphate ester liberates p-nitrophenol, a soluble, yellow chromogen that can be measured spectrophotometrically at 405 nm (ε = 1.83 x 104 M-1cm-1). As the reaction is linear, kinetic assays can be performed. Endpoint determinations can also be performed by stopping the reaction with strong base and measuring the absorbance at 405 nm. This p-NPP preparation is a stable, convenient, liquid concentrate in a proprietary solvent.
p-Nitrophenyl phosphate supplied as a 50X concentrate (0.19 mol/l). For use with p-NPP Substrate Buffer, Cat. No. 487664.
Excellent substrate for alkaline phosphatase-based ELISA assays. Produces yellow soluble end product that can be read at 405 nm to 410 nm. Both kinetic and endpoint determinations can be performed. Supplied as ready-to-use. A ready-to-use diluent is available separately (Cat. No. 487664).
This p-Nitrophenyl Phosphate is validated for use in ELISA.

Application

ELISA

Biochem/physiol Actions

Primary Target
Substrate for alkaline phosphatase-based ELISA assays
Product does not compete with ATP.

Physical form

Supplied as a 50X concentrate.

Preparation Note

Discard if solution turns bright yellow or becomes turbid.

Other Notes

Functionality check performed on the final product in ELISA.



Suggested procedure for use of p-NPP in alkaline phosphatase-based ELISAs:



1. Prepare 1X p-NPP Substrate Solution: To 39 parts dH2O add 1 part 50X p-NPP Concentrate and 10 parts p-NPP Substrate Buffer (Cat. No. 487664). Mix well and store protected from light. This diluted reagent is suitable for up to 10 h after preparation.



Note: Cat. No. 487664 is an AMP (2-amino-2-methyl-1-propanol)-based buffer. Other buffers, e.g. diethanolamine (DEA)-based and 2-amino-2-methyl-1,3-propanediol (AMPD)-based, may also be employed.



2. Complete all required incubations with antibodies and alkaline phosphatase-labeled probes.

3. Wash plate wells at least 4 times with Tris-buffered saline (TBS) containing 0.1% Tween®-20 detergent. DO NOT USE PHOSPHATE BUFFERS; inorganic phosphate is a potent inhibitor of alkaline phosphatase.

4. After the final wash, shake and blot all residual buffer from the wells.

5. Add 100 µl of 1X p-NPP Substrate Solution (prepared in Step 2) to each well and incubate for 15-60 min. (If a kinetic study is desired, readings at 405 nm may be taken at intermediate intervals).



[Note: Optimal incubation times may vary depending on the amount of enzyme present. If color develops too quickly, dilution of the probe, antibody, or alkaline phosphatase-labeled reagent may be required.]



6. Stop the reaction by adding 100 µl 1 N NaOH.

7. Read absorbance at 405 nm.



Note: Variations in time, reagent volumes, and temperature may require further standardization by the user.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4876487655487666
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

form

liquid

form

crystalline solid

form

solid

form

-

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze

storage condition

OK to freeze

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

does not contain

preservative

does not contain

-

does not contain

-

does not contain

-


Still not finding the right product?

Explore all of our products under p-Nitrophenyl Phosphate


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters