Skip to Content
Merck
All Photos(3)

Key Documents

4.80531

Supelco

Sulfuric acid

62%, for the determination of fat in cheese (d 1.52) EMSURE®

Synonym(s):

Sulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

Pricing and availability is not currently available.

Product Name

Sulfuric acid 62% for analysis, for the determination of fat in cheese (d 1.52) EMSURE®

grade

reagent

Quality Level

product line

EMSURE®

form

liquid

concentration

61.5-62.5% H2SO4

density

1.52 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

Looking for similar products? Visit Product Comparison Guide

Application


  • Optimization of Culture Medium for Bio-cementation: Sulfuric acid played a critical role in optimizing the culture medium to enhance the bio-cementation effect, crucial for construction and material science. This study leverages the acid′s properties to improve the structural integrity of bio-based materials (Pan et al., 2024).

  • Sustainable Aerogel Development: A study highlighted the use of sulfuric acid in the production of lignocellulose aerogel aimed at air purification with additional benefits such as thermal insulation and flame retardancy. This research underlines the acid′s versatility in developing advanced materials that are critical for environmental sustainability and public safety (Sun et al., 2024).

Analysis Note

Assay (alkalimetric): 61.5 - 62.5 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service