Skip to Content
Merck
All Photos(1)

Documents

344270

Sigma-Aldrich

Forskolin

from Coleus forskohlii, ≥99% (HPLC), crystalline solid, positive inotropic agent, Calbiochem

Synonym(s):

Forskolin, Coleus forskohlii, 7β-Acetoxy-8,13-epoxy-1α,6β,9α-trihydroxy-labd-14-en-11-one, Colforsin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O7
CAS Number:
Molecular Weight:
410.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Forskolin, Coleus forskohlii,

Assay

≥99% (HPLC)

Quality Level

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C22H34O7/c1-8-19(5)11-14(25)22(27)20(6)13(24)9-10-18(3,4)16(20)15(26)17(28-12(2)23)21(22,7)29-19/h8,13,15-17,24,26-27H,1,9-11H2,2-7H3/t13-,15-,16-,17-,19-,20-,21+,22-/m0/s1

InChI key

OHCQJHSOBUTRHG-KGGHGJDLSA-N

General description

A cardioactive diterpene isolated from the Indian plant Coleus forskolii. At low doses, acts as a positive inotropic agent in dogs, cats, spontaneously hypertensive and normal rats, and isolated guinea pig heart. At higher doses, acts as a hypotensive and vasodilatory agent due to its action as a smooth muscle relaxant. No major side effects are observed at effective doses. Rapid and reversible activator of adenylate cyclase (EC50 = 4 µM) in membranes and intact cells. Does not affect the activity of guanylate cyclase or cyclic nucleotide phophodiesterases. The exact mechanism of forskolin′s positive inotropic effect is unknown but may be related to a cAMP-dependent increase in Na+ permeability, which results in indirect augmentation of Ca2+ release. Inhibits MAP kinase in rat renal mesangial cells (IC50 = 25 µM). Also acts as a Hh pathway antagonist. Shown to inhibit apoptosis in cerebellar granule cells and to induce apoptosis in resting human B-lymphocytes.
The major cell-permeable diterpene isolated from the Indian plant Coleus forskohlii. At low doses, it acts as a positive inotropic agent. At higher doses, it serves as a hypotensive and vasodilatory agent due to its actions as a smooth muscle relaxant. No major side effects are observed at effective doses. Forskolins pharmacological activities are due to its activation of adenylate cyclase (EC50 = 4 µM), resulting in increased cAMP levels. The exact mechanism of forskolins positive inotropic effect is unknown but may be related to a cAMP-dependent increase in Na+ permeability that results in indirect augmentation of Ca2+ release. Inhibits MAP kinase in rat renal mesangial cells (IC50 = 25 µM). Also acts as a Hh pathway antagonist. Shown to inhibit apoptosis in cerebellar granule cells and to induce apoptosis in resting human B lymphocytes.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target IC50: 25 µM inhibiting MAP kinase in rat renal mesangial cells; EC50 = 4 µM for activation of adenylate cyclase

Caution

Ethanol is reported to inhibit forskolin-mediated activation of adenylate cyclase. DMSO is the recommended solvent; at ≤5% DMSO final concentration, little inhibition of the adenylate cyclase activation will occur.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). DMSO stock solutions are stable for up to 4 months at 4°C.

Other Notes

D′Orazio, J.A., et al. 2006. Nature443, 340.
Noveen, A., et al. 1996. Biochem. Biophys. Res. Commun.219, 180.
Galli, C., et al. 1995. J. Neurosci.15, 1172.
Li, X., et al. 1995. Am. J. Physiol.269, C986.
Lomo, J., et al. 1995. J. Immunol.154, 1634.
Uneyama, H., et al. 1993. J. Biol. Chem.268, 168.
Laurenza, A., et al. 1989. Trends Pharmacol. Sci.10, 442.
Adashi, E.Y., and Resnick, C.E. 1986. J. Cell. Biochem.31, 217.
Seamon, K.B., and Daly, J.W. 1986. Adv. Cyclic Nucleotide Protein Phosphorylation Res.20, 1.
Huang, R., et al. 1982. Cyclic Nucleotide Res.8, 385.
Metzger, H., and Lindner, E. 1981. IRCS Med. Sci. Biochem. Cardiovasc. System Pharmacol.9, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rashika Joshi et al.
American journal of physiology. Lung cellular and molecular physiology, 322(1), L162-L173 (2021-12-02)
Most lung development occurs in the context of cyclic stretch. Alteration of the mechanical microenvironment is a common feature of many pulmonary diseases, with congenital diaphragmatic hernia (CDH) and fetal tracheal occlusion (FETO, a therapy for CDH) being extreme examples
Cecile Jacovetti et al.
Molecular metabolism, 84, 101955-101955 (2024-05-05)
The contribution of the mitochondrial electron transfer system to insulin secretion involves more than just energy provision. We identified a small RNA fragment (mt-tRF-LeuTAA) derived from the cleavage of a mitochondrially-encoded tRNA that is conserved between mice and humans. The
Cheng-Guo Wu et al.
Proceedings of the National Academy of Sciences of the United States of America, 121(1), e2310727120-e2310727120 (2023-12-27)
Intrinsically disordered regions (IDR) and short linear motifs (SLiMs) play pivotal roles in the intricate signaling networks governed by phosphatases and kinases. B56δ (encoded by PPP2R5D) is a regulatory subunit of protein phosphatase 2A (PP2A) with long IDRs that harbor
Lara El-Bazzal et al.
Brain : a journal of neurology, 146(5), 1844-1858 (2022-11-01)
Charcot-Marie-Tooth (CMT) disease is one of the most common inherited neurological disorders, affecting either axons from the motor and/or sensory neurons or Schwann cells of the peripheral nervous system (PNS) and caused by more than 100 genes. We previously identified
Ieva Sutkeviciute et al.
Nature chemical biology, 18(3), 272-280 (2021-12-25)
Class B G protein-coupled receptors (GPCRs) are notoriously difficult to target by small molecules because their large orthosteric peptide-binding pocket embedded deep within the transmembrane domain limits the identification and development of nonpeptide small molecule ligands. Using the parathyroid hormone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service