Skip to Content
Merck
All Photos(1)

Documents

324408

Sigma-Aldrich

Anti-Dopamine D₅ Receptor Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

does not contain

preservative

species reactivity

human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DRD5(1816)

Related Categories

General description

Anti-Dopamine D5 Receptor, rabbit polyclonal, recognizes the dopamine D5 receptor in rat brain. Does not cross-react with other dopamine receptors. It is validated for use in ELISA, WB, FC, and IHC.
Rabbit polyclonal antibody supplied as lyophilized, undiluted serum. Recognizes the ~49 kDa D5 dopamine receptor protein.
Recognizes the dopamine D5 receptor in rat brain. Does not cross-react with other dopamine receptors.

Immunogen

Human
a synthetic peptide (LPPGSNGTAYC) corresponding to amino acids 2-10 of human dopamine D₅ receptor, conjugated to carrier protein

Application

ELISA (see comments)

Flow Cytometry (1:100, see application references)

Immunoblotting (1:200)

Immunohistochemistry (1:2500)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Reconstitute the lyophilized antibody with 100 µl PBS containing 10 mg/ml BSA. For further dilution of the antibody use a similar buffer formulation. Following reconstitution, refrigerate (4°C).

Analysis Note

Positive Control
Rat brain

Other Notes

For immunohistochemistry, a 1:2500 dilution of the reconstituted antiserum demonstrated staining of specific cells in various regions of rat brain sections, including Cortex layer 5 and parafascicular nucleus. This antibody has also been reported to work for ELISA. Variables associated with assay conditions will dictate the proper working dilution.
McKenna, F., et al. 2002. J. Neuroimmunol.132, 34.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonis Prosilis et al.
The International journal of developmental biology, 66(1-2-3), 263-267 (2021-12-10)
Even before the first synapses appear, neurotransmitters and their receptors are present in the developing brain, regulating the cell fate of neuronal progenitors in neurogenic niches, such as the lateral ventricle. In particular, dopamine appears to play a pivotal role
Kentaro Ono et al.
Scientific reports, 7(1), 9181-9181 (2017-08-25)
We propose a new mechanism of sensory modulation through cutaneous dopaminergic signalling. We hypothesize that dopaminergic signalling contributes to differential cutaneous sensitivity in darker versus lighter pigmented humans and mouse strains. We show that thermal and mechanical cutaneous sensitivity is
Rajasekharreddy Pala et al.
Nano letters, 19(2), 904-914 (2018-12-26)
Ciliopathies caused by abnormal function of primary cilia include expanding spectrum of kidney, liver, and cardiovascular disorders. There is currently no treatment available for patients with cilia dysfunction. Therefore, we generated and compared two different (metal and polymer) cilia-targeted nanoparticle
Rajasekharreddy Pala et al.
ACS nano, 13(3), 3555-3572 (2019-03-13)
Patients with polycystic kidney disease (PKD) are characterized with uncontrolled hypertension. Hypertension in PKD is a ciliopathy, an abnormal function and/or structure of primary cilia. Primary cilia are cellular organelles with chemo and mechanosensory roles. In the present studies, we
Magdalena Kurnik-Łucka et al.
International journal of molecular sciences, 22(23) (2021-12-11)
an increased prevalence of gastro-duodenal ulceration was described almost sixty years ago as prodromal to idiopathic Parkinson's disease, while duodenal ulcers have been rarely diagnosed in patients with schizophrenia. The cytoprotective role of dopamine in animal models of gastrointestinal ulcerations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service