Skip to Content
Merck
All Photos(1)

Key Documents

219491

Millipore

Cdk/Crk Inhibitor

The Cdk/Crk Inhibitor, also referenced under CAS 784211-09-2, controls the biological activity of Cdk/Crk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk/Crk Inhibitor, 1-(2,6-Dichlorophenyl)-1,5-dihydro-6-((4-(2-hydroxyethoxy)phenyl)methyl)-3-(1-methylethyl)-4H-pyrazolo[3,4-d]pyrimidin-4-one, RGB-286147, Cdk7 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22Cl2N4O3
CAS Number:
Molecular Weight:
473.35
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable pyrazolo-pyrimidinone compound that acts as a potent, selective, and ATP-competitive inhibitor of Cdks (IC50 = 48 nM, 15 nM, 9 nM, 10 nM, 71 nM, and 9 nM for Cdk1/B, Cdk2/E, Cdk3/E, Cdk5/p35, Cdk7/H/MAT1, and Cdk9, respectively). It inhibits Cdk4/D1, Cdk6/D3, and GSK-3β only at much higher concentrations (IC50 = 839 nM, 282 nM, and 754 nM, respectively) and exhibits little activity towards a panel of 60 other kinases. A broad-spectrum growth inhibitor toward tumor cells, but not non-transformed quiescent fibroblasts.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk1/B, Cdk2/E, Cdk3/E, Cdk5/p35, Cdk7/H/MAT1, and Cdk9
Product competes with ATP.
Reversible: no
Target IC50: 48 nM, 15 nM, 9 nM, 10 nM, 71 nM, and 9 nM for Cdk1/B, Cdk2/E, Cdk3/E, Cdk5/p35, Cdk7/H/MAT1, and Cdk9, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Caligiuri, M., et al. 2005. Chem. Biol.12, 1103.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacologic Targeting of TFIIH Suppresses KRAS-Mutant Pancreatic Ductal Adenocarcinoma and Synergizes with TRAIL.
Moser, et al.
Cancer research, 82, 3375-3393 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service