Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352200
NACRES:
NA.41
eCl@ss:
32160405
Form:
liquid
Solubility:
water: soluble
Product Name
Kinase Inhibitor Cocktail, The Inhibitor Cocktail controls the biological activity of kinases. This small molecule/inhibitor is primarily used for Biochemicals applications.
Quality Segment
form
liquid
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
solubility
water: soluble
shipped in
wet ice
Physical form
20mM MOPS, pH 7.2, 25mM β-glycerol phosphate, 5mM EGTA, 1mM sodium orthovanadate, 1mM dithiothreitol.
Analysis Note
Routinely evaluated in kinase assays as an inhibitor.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 20-116 | 04053252401350 |
| THSD1145-3EA | 04061834000297 |