Skip to Content
Merck
All Photos(1)

Key Documents

189400

Sigma-Aldrich

Aurintricarboxylic Acid

A cell-permeable polyanionic, polyaromatic compound used as a powerful inhibitor of cellular processes that are dependent on the formation of protein-nucleic acid complexes.

Synonym(s):

Aurintricarboxylic Acid, ATA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14O9
CAS Number:
Molecular Weight:
422.34
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Quality Level

description

RTECS - GU4790000

Assay

≥85% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

red

solubility

0.1 M NaOH: 35 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C22H16O9.3H3N/c23-16-4-1-10(7-13(16)20(26)27)19(11-2-5-17(24)14(8-11)21(28)29)12-3-6-18(25)15(9-12)22(30)31;;;/h1-10,19,24-25H,(H,26,27)(H,28,29)(H,30,31);3*1H3

InChI key

UDPQMNAHKKTHHI-UHFFFAOYSA-N

General description

A cell-permeable polyanionic, polyaromatic compound used as a powerful inhibitor of cellular processes that are dependent on the formation of protein-nucleic acid complexes. Binds to aFGF and reduces its angiogenic activity. Shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. ATA is a potent inhibitor of DNA topoisomerase II (IC50= 75 nM for the yeast enzyme as measured by relaxation assays) that also acts as a potent inhibitor of angiogenesis. Inhibits von Willebrand factor binding to platelets and reduces glutamate-induced neuronal injury. ATA stimulates the tyrosine phosphorylation of MAP kinases, Shc proteins, phosphatidylinositol 3-kinase, and phospholipase Cγ. Inhibits both major calpain isoforms (IC50 = 22 µM and IC50 = 10 µM for µ-calpain and m-calpain, respectively).
Dye content: ~85%.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA topoisomerase 2
Product does not compete with ATP.
Reversible: no
Target IC50: 75 nM against DNA topoisomerase II

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot, purge with inert gas, and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Lozano, R.M., et al. 1997. Eur. J. Biochem.248, 30.
Benchokroun, Y., et al. 1995. Biochem. Pharmacol.49, 305.
Okada, N., and Koizumi, S. 1995. J. Biol. Chem.270, 16464.
Posner, A., et al. 1995. Biochem. Mol. Biol. Int. 36, 291.
Catchpoole, D.R., et al. 1994. Anticancer Res. 14, 853.
Csernansky, C.A., et al. 1994. J. Neurosci. Res. 38, 101.
Gagliardi, A.R., and Collins, D.C. 1994. Anticancer Res. 14, 475.
Gonzalez, R.G., et al. 1980. Biochemistry19, 4299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bidur Paudel et al.
Nature communications, 15(1), 264-264 (2024-01-19)
Alzheimer's disease (AD) is characterized by progressive neurodegeneration, but the specific events that cause cell death remain poorly understood. Death Induced by Survival gene Elimination (DISE) is a cell death mechanism mediated by short (s) RNAs acting through the RNA-induced
Norie Sugitani et al.
Cell reports, 40(12), 111371-111371 (2022-09-22)
ATR kinase is a central regulator of the DNA damage response (DDR) and cell cycle checkpoints. ATR kinase inhibitors (ATRi's) combine with radiation to generate CD8+ T cell-dependent responses in mouse models of cancer. We show that ATRi's induce cyclin-dependent kinase
Le D He et al.
Bio-protocol, 7(5), e2157-e2157 (2017-03-05)
Drechmeria coniospora is a nematophagous fungus and potential biocontrol agent. It belongs to the Ascomycota. It is related to Hirsutella minnesotensis, another nematophagous fungus but, phylogenetically, it is currently closest to the truffle parasite Tolypocladium ophioglossoides. Together with its natural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service