Skip to Content
Merck
All Photos(1)

Key Documents

17-408

Sigma-Aldrich

EZ-Magna ChIP® A - Chromatin Immunoprecipitation Kit

Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads. Control primers included.

Synonym(s):

Magnetic ChIP Kit, Magnetic Chromatin Immunoprecipitation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.52

Quality Level

manufacturer/tradename

Magna ChIP®

technique(s)

immunoprecipitation (IP): suitable

shipped in

dry ice

General description

Chromatin Immunoprecipitation (ChIP) is a powerful technique for mapping the in vivo distribution of proteins associated with chromosomal DNA. These proteins can be histone subunits and post-translational modifications or other chromatin associated proteins such as transcription factors, chromatin regulators, etc. Additionally, ChIP can be used to identify regions of the genome associated with these proteins, or conversely, to identify proteins associated with a particular region of the genome. ChIP methodology often involves protein-DNA and protein-protein cross-linking, fragmentation of the cross-linked chromatin, and subsequent immunoprecipitation of chromatin with an antibody specific to a target protein. The DNA fragments isolated in complex with the target protein can be identified by a variety of methods including PCR, DNA microarray and DNA sequencing. Standard or quantitative PCR can be performed to verify whether a particular DNA sequence (the gene or region of the genome) is associated with the protein of interest. The combination of ChIP and promoter or genomic tiling microarrays (ChIP-chip) allows genome-wide identification of DNA-binding sites for chromatin-associated proteins with precise resolution. Alternatively, high-throughput sequencing of libraries constructed from immunoprecipitated chromosomal DNA (ChIP-Seq) is a powerful alternative to ChIP-chip in mapping the protein-DNA interactions across mammalian genomes.
Chromatin Immunoprecipitation (ChIP) is an important technique allowing the researcher to analyze in vivo interactions of proteins with genomic DNA. Any chromatin-associated or DNA binding protein can be analyzed with this technique, provided a good antibody to the protein exists. One can measure different proteins localized to a specific region of the genome, or the genome wide distribution of a specific protein. Another powerful application of this technique is to analyze changes in histone modifications that correlate with processes like transcription, mitosis or DNA repair.

Features & Benefits:
Faster: Magnetic protein A beads allow for the entire ChIP protocol to be done in as little as a day! All reagents to process your samples are included - you don′t have to spend valuable time making them.
Easier: Spin columns make DNA purification easier and more reliable - no more messy phenol-chloroform extractions.
Greater Reproducibility: Positive and negative control antibodies and PCR primers are included to help validate your results and to troubleshoot your experiments.

Application

Used to detect/quantify: Protein A

Packaging

Kit capacity: 22 chromatin immunoprecipitation assays

Components

Magnetic Protein A beads

ChIP Dilution Buffer

Low Salt Wash Buffer

High Salt Wash Buffer

LiCl Wash Buffer

TE Buffer

Cell Lysis Buffer

Nuclear Lysis Buffer

ChIP Elution Buffer (w/o Proteinase K)

10X Glycine

10X PBS

Protease Inhibitor Cocktail II

Proteinase K

Control Primers

Anti-Acetyl Histone H3

Normal Rabbit IgG

Spin Filters

Collection Tubes

Bind Reagent A

Wash Reagent B

Elution Reagent C

Physical form

Two boxes containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions. Supplied buffers are sufficient to generate chromatin from up to five 15 cm plates of cultured cells, each plate providing up to 10 chromatin preparations (varies with cell and assay type).

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels. Kit components are stable for 1 year from date of shipment when stored as directed.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Xu et al.
Cellular oncology (Dordrecht), 43(2), 297-310 (2019-12-31)
Long non-coding RNAs (lncRNAs) are known to be frequently dysregulated in many types of human cancer. As yet, however, their roles in colon carcinogenesis have not been fully elucidated. In the current study, we assessed whether lncRNA LINC00858 may be
Lei Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 2703-2714 (2020-01-10)
Diabetic nephropathy (DN) is a frequently occurred microvascular complication associated with type I and type II diabetes mellitus. The participation of long noncoding RNAs (lncRNAs) in diabetes-related microvascular complications has been reported extensively. We attempted to unveil the possible regulatory
Songjun Xu et al.
PloS one, 11(7), e0159623-e0159623 (2016-07-28)
Environmental enrichment (EE) conditions have beneficial effects for reinstating cognitive ability in neuropathological disorders like Alzheimer's disease (AD). While EE benefits involve epigenetic gene control mechanisms that comprise histone acetylation, the histone acetyltransferases (HATs) involved remain largely unknown. Here, we
Characterization of the activation of protein tyrosine phosphatase, receptor-type, Z polypeptide 1 (PTPRZ1) by hypoxia inducible factor-2 alpha.
Wang, V; Davis, DA; Veeranna, RP; Haque, M; Yarchoan, R
Testing null
IRF-1 transcriptionally upregulates PUMA, which mediates the mitochondrial apoptotic pathway in IRF-1-induced apoptosis in cancer cells.
Gao, J; Senthil, M; Ren, B; Yan, J; Xing, Q; Yu, J; Zhang, L; Yim, JH
Cell Death and Differentiation null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service