Skip to Content
Merck
All Photos(5)

Key Documents

07-327

Sigma-Aldrich

Anti-acetyl-Histone H4 (Lys5) Antibody

serum, from rabbit

Synonym(s):

Histone H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys5)

Gene Information

General description

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).

Specificity

This antibody is expected to demonstrate broad species cross-reactivity.
This antibody recognizes the N-terminus of Histone H4 acetylated at Lys5.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of Histone H4 acetylated at Lys5.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected acetyl-Histone H4 in HeLa and A431 cells.

Dot Blot Specificity Analysis: A representative lot detected acetyl-histone H4 (Lys5) peptides, but not unacetylated-histone H4 (Lys4) peptides or other peptides corresponding to modified histones.

Chromatin Immunoprecipitation Analysis: 5 µL from a representative lot immunoprecipitated acetyl-Histone H4 in HeLa S3 chromatin.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-acetyl-Histone H4 (Lys5) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB, ICC, ChIP-seq, ChIP to detect acetyl-Histone H4 (Lys5) also known as H4K5Ac, Histone H4 (acetyl K5).

Quality

Evaluated by Western Blot in HeLa acid extract.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected acetyl-Histone H4 in 10 µg of HeLa acid extract.

Target description

~13 kDa observed

Physical form

Immunodepleted rabbit serum with 0.05% soduim azide and 30% glycerol.
Purified by immunodepletion

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
HeLa acid extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AP-2alpha induces epigenetic silencing of tumor suppressive genes and microsatellite instability in head and neck squamous cell carcinoma.
Bennett, KL; Romigh, T; Eng, C
Testing null
Long-Kuan Ran et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(13), 3372-3382 (2016-02-11)
To characterize the functional role of SIRT6 in hepatocellular carcinoma (HCC). The expression of SIRT6 in 60 paired paraffin-embedded HCC tissues and adjacent nontumoral liver tissues was examined by immunohistochemistry. The expression of SIRT6 in 101 paired frozen HCC tissues
Fei Gao et al.
International journal of nanomedicine, 11, 3859-3874 (2016-08-30)
As an engineered nanomaterial, zinc oxide nanoparticles (ZnO NPs) are used frequently in biological applications and can make contact with human skin. Here, we systematically investigated the effects of ZnO NPs on non-tumorigenic human epidermal keratinocytes, which were used as
Kathryn L Huisinga et al.
Genetics, 202(2), 565-582 (2015-12-19)
Heterochromatin is a common DNA packaging form employed by eukaryotes to constitutively silence transposable elements. Determining which sequences to package as heterochromatin is vital for an organism. Here, we use Drosophila melanogaster to study heterochromatin formation, exploiting position-effect variegation, a
Myriam Rai et al.
PloS one, 5(1), e8825-e8825 (2010-01-26)
Friedreich's ataxia (FRDA), the most common recessive ataxia in Caucasians, is due to severely reduced levels of frataxin, a highly conserved protein, that result from a large GAA triplet repeat expansion within the first intron of the frataxin gene (FXN).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service