Skip to Content
Merck
All Photos(3)

Key Documents

17-10521

Sigma-Aldrich

EZ-Magna NuCLEAR RIP (Cross-Linked) Nuclear RNA-Binding Protein Immunoprecipitation Kit

EZ-Magna Nuclear RIP (Cross-Linked) RNA-Binding Protein Immunoprecipitation Kit is designed for the analysis of chromatin associated RNA such lncRNAs, enhancer RNAs and miRNAs.

Synonym(s):

Magnetic RNA-BP Immunoprecipitation, RNA-Binding Protein Immunoprecipitation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Quality Level

manufacturer/tradename

Magna Nuclear RIP

technique(s)

RIP: suitable
activity assay: suitable (protein interaction)
immunoprecipitation (IP): suitable

shipped in

dry ice

Related Categories

General description

Features and Advantages

  • Generates cross-linked chromatin to allow analysis of a variety of chromatin-associated RNAs
  • Flexible, scalable input requirements: Recover RNA from millions of cells or as few as 5,000 cells
  • Magnetic protein A/G bead blend and optimized buffer system results in low backgrounds and high signal-to-noise ratios
  • Suitable for analysis by RT-qPCR or RIP-seq
  • Complete set of reagents and detailed protocol to enable first-time success


Magna Nuclear RIP Kits are specially designed to allow the discovery and analysis of a variety of chromatin associated RNAs such as long non-coding RNAs, enhancer RNAs and miRNAs . These chromatin-associated RNAs often regulate gene expression and can be analyzed with applications including quantitative reverse transcription polymerase chain reaction (RT-PCR), microarray analysis (RIP-chip) and next generation sequencing (RIP-Seq).

Nuclear RIP can be performed using chromatin that has interactions stabilized by formaldehyde treatment (cross-linked) or chromatin that has not been treated with a cross linking reagent (native). While both of these approaches are similar in that they are designed to recover chromatin associated RNA, the reagents used and the details of the protocol and types of interactions typically detected are different. Cross-linked can capture higher molecular weight complexes in in vivo configurations with possibly lower affinities. In contrast native RIP is expected to recover high affinity, more direct interactions between proteins encoded RNA binding motifs and candidate RNAs. For less well understood proteins and protein complexes often both approaches are used.

The kit described here is for the cross-linked approach. If a native approach is of interest please visit the product page for the Magna Nuclear RIP (Native) Kit, catalogue # 17-10522 or the EZ-Magna Nuclear RIP (Native) Kit, catalogue # 17-10523.

Packaging

Kit capacity: 24 RNA-binding protein immunoprecipitation assays using a cross-linked nuclear lysate

Components

10X Glycine

10X PBS

Nuclei Isolation Buffer

RIP Cross-Linked Lysis Buffer

Protein A/G Magnetic Beads

Nuclear RIP Dilution Buffer

Low Salt Wash Buffer

High Salt Wash Buffer

LiCl Wash Buffer

TE Buffer

RIP Elution Buffer

10% SDS

0.5 M EDTA

DNase I (RNase Free)

DNase I Supplement

DNase I Reaction Buffer

Protease Inhibitor Cocktail III, Animal Free

RNAse Inhibitor

Proteinase K


Control Antibodies and Primers
Normal Mouse IgG Negative Control Antibody

Anti-EZH2 Positive Control Antibody

NEAT1 Positive Control Primers

U1 snRNA Negative Control Primers

Physical form

Two boxes containing key reagents for generation of cross linked nuclear lysates and performance of 24 individual RNA-binding protein immunoprecipitation (RIP) reactions. Plus box containing positive and negative control antibodies and qPCR primers.

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels.
Kit components are stable for 6 months from date of shipment when stored as directed.

Legal Information

NuCLEAR is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10-13 - German Storage Class 10 to 13

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengsi Hu et al.
Journal of cellular and molecular medicine, 21(11), 2732-2747 (2017-04-27)
Metastasis associated lung adenocarcinoma transcript 1(MALAT1) is a long non-coding RNA, broadly expressed in mammalian tissues including kidney and up-regulated in a variety of cancer cells. To date, its functions in podocytes are largely unknown. β-catenin is a key mediator
Jingyi Song et al.
Molecules (Basel, Switzerland), 23(12) (2018-12-14)
Glioblastoma (GBM), the most common type of primary tumor in the central nervous system, is a very aggressive brain tumor with poor prognosis and a high recurrence rate. Increasing evidence suggests that human cytomegalovirus (HCMV) infection is related to GBM
Xiaoli Zhang et al.
iScience, 24(10), 103097-103097 (2021-10-09)
The serine/arginine-rich (SR) family of splicing factors plays important roles in mRNA splicing activation, repression, export, stabilization, and translation. SR-splicing factor 5 (SRSF5) is a glucose-inducible protein that promotes tumor cell growth. However, the functional role of SRSF5 in tissue
Michele Spiniello et al.
Journal of proteome research, 17(9), 3022-3038 (2018-07-05)
RNA-protein interactions are integral to the regulation of gene expression. RNAs have diverse functions and the protein interactomes of individual RNAs vary temporally, spatially, and with physiological context. These factors make the global acquisition of individual RNA-protein interactomes an essential
Michele Spiniello et al.
RNA (New York, N.Y.), 25(10), 1337-1352 (2019-07-13)
Proteins bind mRNA through their entire life cycle from transcription to degradation. We analyzed c-Myc mRNA protein interactors in vivo using the HyPR-MS method to capture the crosslinked mRNA by hybridization and then analyzed the bound proteins using mass spectrometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service