Skip to Content
Merck
All Photos(1)

Key Documents

1.04483

Supelco

ICP Multi element standard solution IX certified reference material

(9 elements in diluted nitric acid), Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

technique(s)

ICP: suitable

matrix

(in diluted nitric acid)

pH

0.3 (20 °C in water)

bp

100 °C/1013 hPa

density

1.05 g/cm3 at 20 °C

application(s)

environmental
food and beverages
personal care
(mining)
(industry & testing)

format

multi-component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.


Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.


Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Packaging

100ml in HD-PE bottle

Analysis Note

As (arsenic): 100 mg/l
Be (beryllium): 100 mg/l
Pb (lead): 100 mg/l
Cd (cadmium): 100 mg/l
Cr (chromium): 100 mg/l
Ni (nickel): 100 mg/l
Hg (mercury): 100 mg/l
Se (selenium): 100 mg/l
Tl (thallium): 100 mg/l

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

Certipur is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service