Skip to Content
MilliporeSigma

1.04498

ICP Multi element standard solution IV certified reference material

(23 elements in diluted nitric acid), Certipur®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Certipur®

technique(s)

ICP: suitable

matrix

(in diluted nitric acid)

pH

1 (20 °C in water)

density

1.09 g/cm3 at 20 °C

application(s)

environmental
food and beverages
pharmaceutical
(Battery testing and Analysis)
(mining)

format

multi-component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.   

Please visit  ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.   

Download your certificate at  https://www.sigmaaldrich.com/US/en/documents-search  to view certified values, including uncertainty and expiry date.

Application

Multielement standard solution IV can be used as a calibration standard in the determination of (ultra-)trace and spectrally interfered elements like Ag, Cd, V, Cr, As and Se in water using sector field inductively coupled plasma mass spectrometry method (ICP-MS).

Packaging

100ml in HD-PE bottle

Analysis Note

Ag (silver): 1000 mg/l
Al (aluminium): 1000 mg/l
B (boron): 1000 mg/l
Ba (barium): 1000 mg/l
Bi (bismuth): 1000 mg/l
Ca (calcium): 1000 mg/l
Cd (cadmium): 1000 mg/l
Co (cobalt): 1000 mg/l
Cr (chromium): 1000 mg/l
Cu (copper): 1000 mg/l
Fe (iron): 1000 mg/l
Ga (gallium): 1000 mg/l
In (indium): 1000 mg/l
K (potassium): 1000 mg/l
Li (lithium): 1000 mg/l
Mg (magnesium): 1000 mg/l
Mn (manganese): 1000 mg/l
Na (sodium): 1000 mg/l
Ni (nickel): 1000 mg/l
Pb (lead): 1000 mg/l
Sr (strontium): 1000 mg/l
Tl (thallium): 1000 mg/l
Zn (zinc): 1000 mg/l

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

For detailed information for our newly launched ICP-Multi element Standards check the flyer: Commitment to Quality: Elevating Certipur® Standards to Certified Reference Material

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

Certipur is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library