Skip to Content
Merck

Skip To

1.03702

Water

suitable for GC/MS, SupraSolv®

Synonym(s):

DI water, Deionized water

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
UNSPSC Code:
41104213
NACRES:
NA.03
MDL number:
Beilstein/REAXYS Number:
2050024
Technique(s):
GC/MS: suitable
Bp:
100 °C (lit.)
Vapor pressure:
23 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Water, for gas chromatography MS SupraSolv®

vapor pressure

23 hPa ( 20 °C)

Quality Segment

product line

SupraSolv®

form

liquid

technique(s)

GC/MS: suitable

impurities

≤30 ppb TOC

evapn. residue

≤5.0 mg/L

color

APHA: ≤10

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

storage temp.

2-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Analysis Note

Identity (IR-spectrum): passes test
Colour: ≤ 10 Hazen
Evaporation residue: ≤ 5.0 mg/l
TOC: ≤ 30 ppb

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.005770061214211
technique(s)

GC/MS: suitable

technique(s)

GC-HS: suitable

technique(s)

ion chromatography: suitable

technique(s)

immunofluorescence: suitable

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

refractive index

n20/D 1.34 (lit.)

refractive index

n20/D 1.34 (lit.)

refractive index

n20/D 1.34 (lit.)

refractive index

n20/D 1.34 (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

pH

7 (20 °C)

pH

7 (20 °C)

pH

-

pH

6.0-8.0(-25 °C77 °F)


Still not finding the right product?

Explore all of our products under Water


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Do you know the resistivity value @ 25°C?

    1 answer
    1. Unfortunately, resistivity is not a tested parameter for this product. It would need to be determined empirically.

      Helpful?

Reviews

No rating value

Active Filters