Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
Water, for headspace gas chromatography SupraSolv®
Quality Segment
agency
according to Ph. Eur., according to USP
vapor pressure
23 hPa ( 20 °C)
product line
SupraSolv®
form
liquid
technique(s)
GC-HS: suitable
impurities
≤1 μg/g Every residual solvent of class 1 acc. to ICH, ≤10 μg/g Every residual solvent of class 2 acc. to ICH, ≤50 μg/g Every residual solvent of class 3 acc. to ICH
evapn. residue
≤5 mg/L
refractive index
n20/D 1.34 (lit.)
pH
7 (20 °C)
bp
100 °C (lit.)
mp
0 °C (lit.)
density
1.000 g/mL at 3.98 °C (lit.)
storage temp.
2-30°C
SMILES string
O
InChI
1S/H2O/h1H2
InChI key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
General description
Application
- Novel Insights into Sb(III) Oxidation and Immobilization during Ferrous Iron Oxygenation: The Overlooked Roles of Singlet Oxygen and Fe (oxyhydr)oxides Formation.: Discusses the impact of water chemistry on the oxidation processes of antimony and iron, contributing to environmental chemistry and pollution control (Wang et al., 2024).
- Exposure And Health Risk Assessment Of Aflatoxins In Hot Red Pepper Marketed In North Shewa Zone, Oromia Region, Ethiopia.: Assesses the role of water in transporting and dissolving aflatoxins, with implications for food safety and analytical methodologies in developing countries (Alemu Degefe et al., 2024).
Analysis Note
GC/HS
Every residual solvent of class 1 acc. to ICH: ≤ 1 µg/g
Every residual solvent of class 2 acc. to ICH: ≤ 10 µg/g
Every residual solvent of class 3 acc. to ICH: ≤ 50 µg/g
Suitable as solvent for the analysis of residual solvents of ICH classes 1, 2 and 3 according to Ph Eur and USP.
Formic acid, acetic acid, N,N-dimethylacetamide, N,N-dimethy lformamide and dimethyl sulfoxide are not detected.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 200 |
| technique(s) GC-HS: suitable | technique(s) gas chromatography (GC): suitable | technique(s) HPLC: suitable, LC/MS: suitable | technique(s) HPLC: suitable, LC/MS: suitable, UHPLC: suitable |
| form liquid | form liquid | form liquid | form liquid |
| refractive index n20/D 1.34 (lit.) | refractive index n20/D 1.34 (lit.) | refractive index n20/D 1.34 (lit.) | refractive index n20/D 1.34 (lit.) |
| mp 0 °C (lit.) | mp 0 °C (lit.) | mp 0 °C (lit.) | mp 0 °C (lit.), 0.0 °C (32.0 °F) |
| bp 100 °C (lit.) | bp 100 °C (lit.) | bp 100 °C (lit.) | bp 100 °C (lit.) |
Still not finding the right product?
Explore all of our products under Water
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
