Skip to Content
Merck

1.02143

SAFC

Calcium phosphate

dried, EMPROVE® ESSENTIAL, Ph. Eur., BP, E 341 (iii)

Pharma Manufacturing

Synonym(s):

Calcium phosphate, penta-Calcium hydroxide triphosphate, Calcium phosphate tribasic, Tricalcium orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca3O8P2
CAS Number:
Molecular Weight:
310.18
MDL number:
UNSPSC Code:
12352302
E Number:
E341 (iii)
EC Index Number:
231-840-8
Pricing and availability is not currently available.

Agency

BP
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

6-8 (20 °C, 50 g/L in H2O, suspension)

mp

1670 °C

density

3.14 g/cm3 at 20 °C

application(s)

pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[Ca+2].[Ca+2].[Ca+2].[P](=O)([O-])([O-])[O-].[P](=O)([O-])([O-])[O-]

InChI

1S/3Ca.2H3O4P/c;;;2*1-5(2,3)4/h;;;2*(H3,1,2,3,4)/q3*+2;;/p-6

InChI key

QORWJWZARLRLPR-UHFFFAOYSA-H

General description

To address challenges of solid formulation manufacturing, we have developed a comprehensive product portfolio to meet with your solid dosage needs in mind. Although solid dose formulations are one of the the most established dosage forms in the pharmaceutical market, the complexity of manufacturing final drug products is becoming more and more challenging. API stability, release kinetics and bioavailability limitations, such as low API solubility, are just a few of many critical obstacles on the way to the market.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Calcium phosphate is a commonly used ingredient in both food and pharmaceutics. In pharmaceutical applications, calcium phosphate is primarily used as a filler/binder.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service