Skip to Content
Merck

07-681

Anti-phospho-AMPK α (Thr172) Antibody

Upstate®, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human, rat
Application:
WB
Citations:
32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr172)

Gene Information

human ... PRKAA1(5562)

General description

Mr 63 kDa

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids surrounding phosphorylated Thr172 of human AMP-activated protein kinase alpha 2 (AMPKα2)

Application

Anti-phospho-AMPK α (Thr172) Antibody is an antibody against phospho-AMPK α (Thr172) for use in WB.

Biochem/physiol Actions

Recognizes phospho-AMPKα.

Physical form

Format: Purified

Analysis Note

Control
Serum starved HEK293 cells
routinely evaluated by immunoblot on RIPA lysates from overnight starved HEK293 cells and partially purified rat liver AMPK

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



B E Kemp et al.
Biochemical Society transactions, 31(Pt 1), 162-168 (2003-01-28)
The AMP-activated protein kinase (AMPK) is a metabolic-stress-sensing protein kinase that regulates metabolism in response to energy demand and supply by directly phosphorylating rate-limiting enzymes in metabolic pathways as well as controlling gene expression.
Tue L Nielsen et al.
Molecular genetics and metabolism, 123(1), 21-27 (2017-11-28)
McArdle disease (glycogen storage disease type V) is an inborn error of skeletal muscle metabolism, which affects glycogen phosphorylase (myophosphorylase) activity leading to an inability to break down glycogen. Patients with McArdle disease are exercise intolerant, as muscle glycogen-derived glucose
Mathias Fasshauer et al.
Biochimie, 86(11), 779-784 (2004-12-14)
Several adipocyte-secreted factors have been demonstrated to potentially link obesity, insulin resistance, and cardiovascular disease. Among those, adiponectin is an insulin-sensitizing and anti-inflammatory adipokine, concentrations of which are decreased in obesity-associated metabolic and vascular disorders. Recently, two adiponectin receptors (AdipoR)