Skip to Content
Merck
All Photos(1)

Documents

07-250

Sigma-Aldrich

Anti-PP2A Antibody, A subunit

Upstate®, from rabbit

Synonym(s):

Anti-MRD36, Anti-PP2AA, Anti-PP2AAALPHA, Anti-PR65A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, Xenopus, pig, rat

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PPP2R1A(5518)

Specificity

PP2A A subunit, α isoform

Immunogen

KLH-conjugated, synthetic peptide of 13 amino acids (MAAADGDDSLY-Ahx-C), containing amino acids 1-11 of human PP2A, A subunit. Ahx is aminohexanoic acid. The immunizing sequence is conserved in human, mouse, rat, porcine and Xenopus.

Application

Anti-PP2A Antibody, A subunit detects level of PP2A & has been published & validated for use in IP & WB.
Research Sub Category
Synapse & Synaptic Biology

Neurodegenerative Diseases

Quality

Routinely evaluated by immunoblot on RIPA lysates from 3T3/A31 cells.

Target description

63-65kDa

Physical form

100mM triethylamine, 100mM sodium phosphate, 0.02% sodium azide before the addition of glycerol to 30%.

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taha Rehmani et al.
International journal of molecular sciences, 25(5) (2024-03-13)
The slmap gene is alternatively spliced to generate many isoforms that are abundant in developing myocardium. The largest protein isoform SLMAP3 is ubiquitously expressed and has been linked to cardiomyopathy, Brugada syndrome and Hippo signaling. To examine any role in
Rachana Patel et al.
The Journal of clinical investigation, 123(3), 1157-1175 (2013-02-26)
Concurrent activation of RAS/ERK and PI3K/AKT pathways is implicated in prostate cancer progression. The negative regulators of these pathways, including sprouty2 (SPRY2), protein phosphatase 2A (PP2A), and phosphatase and tensin homolog (PTEN), are commonly inactivated in prostate cancer. The molecular
R Ruediger et al.
Oncogene, 20(1), 10-15 (2001-03-13)
The A subunit of protein phosphatase 2A (PP2A) consists of 15 nonidentical repeats. The catalytic C subunit binds to C-terminal repeats 11 - 15 and regulatory B subunits bind to N-terminal repeats 1 - 10. Recently, four cancer-associated mutants of
Sonia Chelouah et al.
Cancers, 10(1) (2018-01-06)
The Epstein-Barr virus nuclear antigen leader protein (EBNA-LP) acts as a co-activator of EBNA-2, a transcriptional activator essential for Epstein-Barr virus (EBV)-induced B-cell transformation. Burkitt's lymphoma (BL) cells harboring a mutant EBV strain that lacks both the EBNA-2 gene and
G A Calin et al.
Oncogene, 19(9), 1191-1195 (2000-03-14)
The phosphatase 2A (PP2A) is one of the major cellular serine-threonine phosphatases. It was recently shown that the gene encoding for the beta isoform of its subunit A, PPP2R1B, is altered in human lung and colorectal carcinomas, suggesting a role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service