Skip to Content
Merck
All Photos(1)

Key Documents

05-1050X

Sigma-Aldrich

Anti-Phosphotyrosine Antibody, 4G10® Platinum

clone 4G10, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4G10, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

General description

The development of the anti-phosphotyrosine, clone 4G10 in 1989 was a monumental discovery for researchers. 4G10 was the first and is the best single monoclonal antibody for the detection tyrosine phosphorylation. 4G10 is well known for its sensitivity and its ability to detect multiple tyrosine phosphorylations on numerous substrates. It has been validated by thousands of scientific and medical researchers in virtually every application and tyrosine target over the past 2 decades. To improve on something that hundred have tried and no one has succeeded, we pooled 4G10 with the next most highly regarded anti-phosphotyrosine, clone PY20 to make 4G10 Platinum. PY20 itself is a very poor substitute for 4G10, but its additive effect allow for a greater level of detection on more substrates that even 4G10 alone was not capable of.

Specificity

Tyrosine-phosphorylated proteins from all species

Immunogen

4G10 used Phosphotyramine coupled to KLH and PY20 used phosphotyrosine conjugated to carrier protein.

Application

Anti-Phosphotyrosine Antibody, 4G10 Platinum is an antibody against Phosphotyrosine for use in WB, IF, FC, IH, IP, IC, ELISA.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Quality

Routinely evaluated by immunoblot on a modified RIPA lysate from EGF-treated human A431 carcinoma cells

Target description

Dependent upon the molecular weight of the tyrosine phosphorylated protein being detected.

Physical form

A proprietary mixture of protein G purified mouse monoclonals 4G10 (IgG2bκ) and PY20 (IgG2b) in 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide.
Format: Purified
Protein G-Sepharose Chromatography

Storage and Stability

2 years at 2-8°C from date of shipment.
NOTE: DO NOT FREEZE.
For maximum recovery of the product, centrifuge the original vial prior to removing the cap. If the product has accidentally been frozen and thawed, spin it at 13,000 x g for 10 minutes at 2-8°C. Save the supernatant for application.

Analysis Note

Control
Positive Antigen Control: Catalog #12-302, EGF-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein expression signatures for inhibition of epidermal growth factor receptor-mediated signaling.
Myers, MV; Manning, HC; Coffey, RJ; Liebler, DC
Molecular and Cellular Proteomics null
Chun-Jung Lin et al.
Evidence-based complementary and alternative medicine : eCAM, 2013, 682418-682418 (2013-02-23)
The bacterial pathogen Helicobacter pylori (Hp) is the leading risk factor for the development of gastric cancer. Hp virulence factor, cytotoxin-associated gene A (CagA) interacted with cholesterol-enriched microdomains and leads to induction of inflammation in gastric epithelial cells (AGS). In
Inmaculada Cuchillo-Ibañez et al.
Scientific reports, 6, 31646-31646 (2016-08-18)
Reelin is a signaling protein that plays a crucial role in synaptic function, which expression is influenced by β-amyloid (Aβ). We show that Reelin and Aβ oligomers co-immunoprecipitated in human brain extracts and were present in the same size-exclusion chromatography
The Kaposi's sarcoma-associated herpesvirus G protein-coupled receptor contains an immunoreceptor tyrosine-based inhibitory motif that activates Shp2.
Philpott, N; Bakken, T; Pennell, C; Chen, L; Wu, J; Cannon, M
Journal of virology null
Olga Samuilova et al.
Journal of virology, 87(8), 4313-4321 (2013-02-01)
Functions of viral proteins can be regulated through phosphorylation by serine/threonine kinases in plants, but little is known about the involvement of tyrosine kinases in plant virus infection. In this study, TGBp3, one of the three movement proteins encoded by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service