Skip to Content
Merck
All Photos(1)

Key Documents

V-011

Supelco

Retinol solution

100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®

Synonym(s):

Retinol solution, 3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexenyl)-2,4,6,8-nonatetraen-1-ol, Axerophthol, Vitamin A alcohol solution, Vitamin A1, all-trans-Retinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O
CAS Number:
Molecular Weight:
286.45
Beilstein:
403040
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

reference material

Quality Level

form

liquid

feature

(Snap-N-Shoot®)
(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT)

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

CC(=C/CO)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H30O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-13,21H,7,10,14-15H2,1-5H3/b9-6+,12-11+,16-8+,17-13+

InChI key

FPIPGXGPPPQFEQ-OVSJKPMPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Enhancing cell culture systems: Research highlights the potential of Retinol as an essential component in cell culture systems, enhancing cell proliferation and differentiation. This supports its use in high-purity Retinol cell culture supplement applications (Ke et al., 2024).
  • Stabilized Retinol in dermatological applications: A study on topical 0.1% stabilized Bioactive Retinol demonstrates its efficacy and tolerability for treating photoaging, indicating its potential as a high-purity Retinol reagent for skin care research (Farris et al., 2024).
  • Drug-nutrient interaction research: Investigation into the interaction between Retinol and pharmaceuticals, like Acetylsalicylic Acid and Warfarin, provides insights into its role in vitamin-based assays, contributing to safer pharmacological studies involving Retinol-based assays (Renaud et al., 2024).
  • Assessment of Retinol signaling pathways: An in silico and in vitro approach examines natural ligands for receptors influenced by Retinol, aiding in the study of Retinol signaling pathways and its application as a research reagent in environmental safety (Choi et al., 2024).

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarmila Mazumder et al.
Lancet (London, England), 385(9975), 1333-1342 (2014-12-17)
Vitamin A supplementation in children aged 6 months to 5 years has been shown to reduce mortality. The efficacy of neonatal supplementation with vitamin A to reduce mortality in the first 6 months of life is plausible but not established.
Gretchen A Stevens et al.
The Lancet. Global health, 3(9), e528-e536 (2015-08-16)
Vitamin A deficiency is a risk factor for blindness and for mortality from measles and diarrhoea in children aged 6-59 months. We aimed to estimate trends in the prevalence of vitamin A deficiency between 1991 and 2013 and its mortality
Malay Rana et al.
Journal of clinical pathology, 67(7), 605-608 (2014-04-09)
Determine the prevalence of fat-soluble vitamin deficiency in children with cystic fibrosis (CF) aged ≤18 years in New South Wales (NSW), Australia, from 2007 to 2010. A retrospective analysis of fat-soluble vitamin levels in children aged ≤18 years who lived in NSW
Emilie Macé et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(1), 7-16 (2014-08-07)
Most inherited retinal dystrophies display progressive photoreceptor cell degeneration leading to severe visual impairment. Optogenetic reactivation of retinal neurons mediated by adeno-associated virus (AAV) gene therapy has the potential to restore vision regardless of patient-specific mutations. The challenge for clinical
Marieke R Beijer et al.
Immunology, 142(1), 39-45 (2014-05-21)
Vitamin A and its active metabolite retinoic acid are essential for the development and function of many tissues including the immune system. The induction of mucosal homing receptors on T and B cells by mucosal dendritic cells (DC) depends on

Protocols

Separation of δ-Tocopherol, analytical standard; Retinyl acetate; (+)-γ-Tocopherol; α-Tocopherol, ≥95.5%; Retinol; Vitamin A (acetate), meets USP testing specifications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service