Skip to Content
Merck
All Photos(1)

Documents

V-007

Supelco

O-Desmethylvenlafaxine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NO2
CAS Number:
Molecular Weight:
263.38
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC=C(C(CN(C)C)C2(O)CCCCC2)C=C1

InChI

1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3

InChI key

KYYIDSXMWOZKMP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

An analytical reference standard applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as urine drug testing, forensic analysis, or clinical toxicology. Also known as desvenlafaxine, O-desmethylvenlafaxine is an SNRI antidepressant marketed under the trade name Pristiq® for the treatment of depression. O-desmethylvenlafaxine is also a major urinary metabolite of venlafaxine, an SNRI antidepressant sold as Effexor® or Efexor and used to treat major depressive disorder, generalized anxiety disorder, and other anxiety disorders associated with depression.

Application


  • Drug Stability in Pharmaceutical Formulations: Another study focused on the physical stability and solubility of O-Desmethylvenlafaxine in various pharmaceutical formulations, contributing to better drug delivery systems and enhanced therapeutic efficacy (Wang et al., 2020).

  • Monitoring Drug Precursors in Advanced Treatment Processes: Research monitored N-nitrosodimethylamine (NDMA) precursors throughout membrane-based advanced wastewater treatment processes, considering the behavior of compounds like O-Desmethylvenlafaxine. This study aids in improving wastewater treatment strategies to mitigate pharmaceutical pollutants (Roccaro et al., 2020).

  • Chiral Drug Analysis in Environmental Waters: A study developed a dual enantioselective LC-MS/MS method for analyzing chiral drugs in surface water, including O-Desmethylvenlafaxine. This method helps in understanding the environmental fate and behavior of chiral pharmaceuticals, enhancing environmental risk assessments (Coelho et al., 2019).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Effexor is a registered trademark of American Home Product Corp.
Pristiq is a registered trademark of Wyeth LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola C Rúa-Gómez et al.
Chemosphere, 90(6), 1952-1959 (2012-11-20)
The photodegradation and biotic transformation of the pharmaceuticals lidocaine (LDC), tramadol (TRA) and venlafaxine (VEN), and of the metabolites O-desmethyltramadol (ODT) and O-desmethylvenlafaxine (ODV) in the aquatic environmental have been investigated. Photodegradation experiments were carried out using a medium pressure
Thomas Guttuso
Maturitas, 72(1), 6-12 (2012-03-02)
Although many non-hormonal compounds have shown statistically significant benefit over placebo in hot flash randomized controlled trials (RCTs), these studies have varied considerably in basic methodology making it challenging to deduce which compounds have the greatest potential to provide clinically
David F Archer et al.
Menopause (New York, N.Y.), 20(1), 47-56 (2012-12-26)
A previous trial of the serotonin-norepinephrine reuptake inhibitor desvenlafaxine (administered as desvenlafaxine succinate) raised concerns on potential serious cardiovascular and hepatic events. The current study was designed to estimate these events in desvenlafaxine versus placebo in a larger population followed
S Unterecker et al.
Pharmacopsychiatry, 45(6), 229-235 (2012-03-20)
Venlafaxine (VEN) is a modern antidepressant which exerts both serotonin and norepinephrine reuptake inhibition. In this study we examined the influence of age, sex, smoking, and co-medication on serum levels of VEN and its metabolite O-desmethylvenlafaxine (ODVEN) in patients treated
Christopher J A Neil et al.
Heart, lung & circulation, 21(4), 203-205 (2012-01-31)
Tako-Tsubo cardiomyopathy (TTC) occurs particularly in post-menopausal women, being precipitated in many cases by severe emotional stress. We describe six patients in whom TTC occurred in association with therapeutic ingestion or overdose of the serotonin/noradrenaline reuptake inhibitor venlafaxine, or its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service