Skip to Content
Merck
All Photos(1)

Key Documents

S-906

Supelco

Stanozolol solution

1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32N2O
CAS Number:
Molecular Weight:
328.49
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in 1,2-dimethoxyethane

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@]1(O)CCC2C3CC[C@H]4Cc5[nH]ncc5C[C@]4(C)C3CC[C@]12C

InChI

1S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24H,4-11H2,1-3H3,(H,22,23)/t14-,15?,16?,17?,19-,20-,21-/m0/s1

InChI key

LKAJKIOFIWVMDJ-LQMCTMSCSA-N

General description

An analytical reference standard applicable for use in sports testing, urine drug testing, or forensic analysis by LC-MS/MS or GC/MS. Stanozolol is used as a treatment for osteoporosis and steroid-deficiency, but has also been abused in sports as a performance-enhancing drug.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An unusual cause of leg ulcerations.
Manuela L Moscaliuc et al.
The American journal of medicine, 124(4), e3-e4 (2011-03-26)
Moritz A Konerding et al.
International journal of molecular medicine, 30(2), 271-276 (2012-05-23)
Despite numerous advances and improvements in surgical techniques the incidence of incisional hernias after laparotomy remains high. The aim of this study was to investigate possible effects of single application of ascorbic acid, stanozolol, a synthetic anabolic steroid, copper peptide
Laxmi S Inamdar Doddamani et al.
The Journal of steroid biochemistry and molecular biology, 129(3-5), 172-178 (2011-12-03)
The abuse of anabolic-androgenic steroids (AAS) for improved physical performance is associated with many deleterious effects. The present study aims to evaluate the short-term effect of an AAS compound stanozolol, on lipoprotein profile, granulopoiesis and immune response in adult female
Adele Chimento et al.
Journal of cellular physiology, 227(5), 2079-2088 (2011-07-20)
Several substances such as anabolic androgenic steroids (AAS), peptide hormones like insulin-like growth factor-I (IGF-I), aromatase inhibitors and estrogen antagonists are offered via the Internet, and are assumed without considering the potential deleterious effects that can be caused by their
Shun-Ye Zhu et al.
Journal of pediatric endocrinology & metabolism : JPEM, 24(5-6), 275-281 (2011-08-10)
Improving the final adult height is one of the most important aims for treatment of central precocious puberty. Stanozolol (ST) is a synthetic derivative of androgen. In this study, we investigated the effects and the mechanisms of ST on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service