Skip to Content
Merck
All Photos(1)

Key Documents

M-906

Supelco

17α-Methyltestosterone solution

1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

17α-Methyl-4-androsten-17β-ol-3-one solution, 17β-Hydroxy-17α-methyl-4-androsten-3-one solution, Mesterone solution, Methyltestosterone solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Beilstein:
2057425
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in 1,2-dimethoxyethane

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C20H30O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h12,15-17,22H,4-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1

InChI key

GCKMFJBGXUYNAG-HLXURNFRSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

17α-Methyltestosterone is an androgenic steroid used in the treatment of testosterone deficiency. 17a-Methyltestosterone is abused in sports as a performance-enhancing drug. This analytical reference standard is applicable for use in sports testing, endocrinology, urine drug testing, or forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atul K Singh
General and comparative endocrinology, 181, 146-155 (2012-10-16)
Of the techniques available to mass-produce a sterile or monosex population, the hormonal induction of sex reversal is the most widely practiced. This paper presents the synthesis of works done on sex reversal using 17α methyltestosterone (17αMT), 17β estradiol, non-steroidal
C Gómez et al.
Steroids, 78(1), 44-52 (2012-11-07)
Methyltestosterone (MT) is one of the most frequently detected anabolic androgenic steroids in doping control analysis. MT misuse is commonly detected by the identification of its two main metabolites excreted as glucuronide conjugates, 17α-methyl-5α-androstan-3α,17β-diol and 17α-methyl-5β-androstan-3α,17β-diol. The detection of these
Anne S Mortensen et al.
Chemosphere, 83(8), 1035-1044 (2011-03-01)
In the present study, groups of juvenile Atlantic salmon (Salmo salar) were fed gelatine capsules containing fish-food spiked with PFOA or PFOS (0.2 mg kg(-1) fish) and solvent (methanol). The capsules were given at days 0, 3 and 6. Blood
Yu Shi et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 163(1), 43-50 (2012-05-15)
Androgens play a crucial role in sex differentiation, sexual maturation, and spermatogenesis in vertebrates. The action of androgens is mediated via androgen receptors (ARs). The present study reports the cloning of the cDNA sequence of the ar in the orange-spotted
Supreeda Homklin et al.
Journal of hazardous materials, 221-222, 35-44 (2012-05-01)
17α-Methyltestosterone (MT), a synthetic anabolic androgenic steroid, is widely used in aquafarming for the production of an all male fish population such as Nile tilapia. This study isolated, identified and characterized MT-degrading bacteria in the sediment and water from a

Protocols

The conversion of clinical methods to LC/MS/MS offers advantages; however, is accompanied by a few limitations, notably interference effects from the endogenous sample matrix.

The conversion of clinical methods to LC/MS/MS offers advantages; however, is accompanied by a few limitations, notably interference effects from the endogenous sample matrix.

The conversion of clinical methods to LC/MS/MS offers advantages; however, is accompanied by a few limitations, notably interference effects from the endogenous sample matrix.

The conversion of clinical methods to LC/MS/MS offers advantages; however, is accompanied by a few limitations, notably interference effects from the endogenous sample matrix.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service