Skip to Content
Merck
All Photos(1)

Documents

N-031

Supelco

Norfentanyl oxalate solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O · C2H2O4
CAS Number:
Molecular Weight:
322.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC(=O)C(O)=O.CCC(=O)N(C1CCNCC1)c2ccccc2

InChI

1S/C14H20N2O.C2H2O4/c1-2-14(17)16(12-6-4-3-5-7-12)13-8-10-15-11-9-13;3-1(4)2(5)6/h3-7,13,15H,2,8-11H2,1H3;(H,3,4)(H,5,6)

InChI key

OLFNESLXXQCODF-UHFFFAOYSA-N

General description

Norfentanyl is a major urinary metabolite of fentanyl, a potent synthetic analgesic sold under various trade names including Sublimaze, Duragesic®, Actiq®, and Fentora®. This Certified Spiking Solution® is suitable for numerous GC/MS or LC/MS testing applications from clinical toxicology and forensic analysis to urine drug testing and pain prescription monitoring.

Legal Information

Actiq is a registered trademark of Anesta Corp.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Duragesic is a registered trademark of Johnson & Johnson
Fentora is a registered trademark of Cima Labs, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Resp. Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vera Coopman et al.
Forensic science international, 169(2-3), 223-227 (2006-05-03)
Fentanyl is a potent synthetic narcotic analgesic administered in the form of a transdermal patch for the management of chronic pain. A 78-year-old woman with a history of cancer was found dead in bed. She was lying on her back.
Henry J Carson et al.
Legal medicine (Tokyo, Japan), 12(3), 157-159 (2010-04-07)
We recently encountered a subject who died from an uncommon misuse of a fentanyl transdermal patch, chewing, followed by complications of aspiration of the patch. We report this case to alert medical examiners to the troubling trend of increased fentanyl
Ming Jin et al.
Journal of analytical toxicology, 29(7), 590-598 (2006-01-20)
Pharmacogenomics, the study on genetic contributions to drug action may help in certifying fentanyl toxicity. Fentanyl is used clinically as an adjunct to surgical anesthesia and for chronic pain management. Its toxicity may be partially due to cytochrome P450 (CYP)
D E Feierman et al.
Drug metabolism and disposition: the biological fate of chemicals, 24(9), 932-939 (1996-09-01)
The microsomal metabolism of fentanyl, a synthetic opioid commonly used in anesthesia, was investigated in human liver. Incubation of fentanyl with human hepatic microsomes fortified with NADPH resulted in the formation of a single major metabolite, namely norfentanyl, as determined
A K Valaer et al.
Journal of chromatographic science, 35(10), 461-466 (1997-10-23)
A sensitive, specific urinary assay for fentanyl, sufentanil, and alfentanil based on their N-dealkylated metabolites is described. Norfentanyl, norsufentanil-noralfentanil, and 2H5-norfentanyl are synthesized and characterized by standard analytical techniques. Derivatization of these secondary amines to yield the pentafluorobenzamides produces stable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service