Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Narcotic Licence Schedule A (Switzerland)
concentration
1.0 mg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
General description
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant® | description 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant® | description 100 μg/mL in methanol (as a free base), certified reference material, ampule of 0.5 mL, Cerilliant® | description 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant® |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| format single component solution | format single component solution | format single component solution | format single component solution |
| application(s) forensics and toxicology | application(s) forensics and toxicology | application(s) forensics and toxicology | application(s) forensics and toxicology |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| drug control Narcotic Licence Schedule A (Switzerland) | drug control Narcotic Licence Schedule A (Switzerland) | drug control - | drug control Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland |
| manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






